Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SFRP1 Antibodies

          Invitrogen

          SFRP1 Polyclonal Antibody

          1 Published Figure
          1 Reference
          View all (21) SFRP1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SFRP1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (1)
          SFRP1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SFRP1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SFRP1 Antibody (PA5-67713) in WB

          Western Blot analysis of SFRP1 was performed using lysates from K562 cells. The membrane was probed with SFRP1 Polyclonal Antibody (Product # PA5-67713) using a dilution of 1:1,000 (4°C). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SFRP1 Antibody in Western Blot (WB)
          SFRP1 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          SFRP1 Polyclonal Antibody

          Product Details

          PA5-67713

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:3,000
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthesized peptide derived from human SFRP1.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2691691

          Product Specific Information

          Predicted to react with Mouse and Rat samples.

          Target Information

          Secreted frizzled-related protein 1 (SFRP1) is a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. SFRP1 and SFRP5 may be involved in determining the polarity of photoreceptor cells in the retina. SFRP1 is expressed in several human tissues, with the highest levels in heart.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: frizzled-related protein; FRP; SARP-2; secreted apoptosis-related protein 2; secreted frizzled-related sequence protein 1; sFRP-1; similar to frizzled-like receptor, extracellular protein; unnamed protein product

          View more View less

          Gene Aliases: 2210415K03Rik; FRP; FRP-1; FRP1; FrzA; SARP2; sFRP-1

          View more View less

          UniProt ID: (Human) Q8N474, (Mouse) Q8C4U3

          View more View less

          Entrez Gene ID: (Human) 6422, (Rat) 84402, (Mouse) 20377

          View more View less

          Function(s)
          cysteine-type endopeptidase activity frizzled binding protein binding heparin binding Wnt-protein binding identical protein binding
          Process(es)
          osteoblast differentiation ureteric bud development positive regulation of cell-matrix adhesion hematopoietic progenitor cell differentiation regionalization positive regulation of cell proliferation negative regulation of cell proliferation cellular response to starvation response to xenobiotic stimulus dorsal/ventral axis specification regulation of cell cycle process negative regulation of gene expression negative regulation of epithelial to mesenchymal transition regulation of neuron projection development Wnt signaling pathway hemopoiesis positive regulation of Wnt signaling pathway negative regulation of Wnt signaling pathway negative regulation of ossification positive regulation of cell growth negative regulation of cell growth negative regulation of cell migration negative regulation of osteoblast proliferation non-canonical Wnt signaling pathway positive regulation of apoptotic process negative regulation of apoptotic process negative regulation of B cell differentiation positive regulation of fat cell differentiation regulation of angiogenesis positive regulation of smoothened signaling pathway negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated negative regulation of bone remodeling negative regulation of fibroblast proliferation stem cell differentiation negative regulation of epithelial cell proliferation negative regulation of peptidyl-tyrosine phosphorylation positive regulation of stress fiber assembly positive regulation of focal adhesion assembly canonical Wnt signaling pathway hematopoietic stem cell differentiation negative regulation of androgen receptor signaling pathway cellular response to vitamin D cellular response to interleukin-1 cellular response to tumor necrosis factor cellular response to prostaglandin E stimulus cellular response to estrogen stimulus cellular response to estradiol stimulus cellular response to hypoxia cellular response to X-ray dopaminergic neuron differentiation cellular response to transforming growth factor beta stimulus cellular response to BMP stimulus negative regulation of canonical Wnt signaling pathway positive regulation of canonical Wnt signaling pathway positive regulation of extrinsic apoptotic signaling pathway via death domain receptors regulation of midbrain dopaminergic neuron differentiation positive regulation of non-canonical Wnt signaling pathway negative regulation of fibroblast apoptotic process positive regulation of fibroblast apoptotic process positive regulation of extrinsic apoptotic signaling pathway somitogenesis neural tube closure male gonad development female gonad development anterior/posterior pattern specification neural crest cell fate commitment neural tube development regulation of ossification negative regulation of BMP signaling pathway developmental process somatic stem cell population maintenance negative regulation of JUN kinase activity negative regulation of osteoblast differentiation negative regulation of osteoclast differentiation development of primary male sexual characteristics digestive tract morphogenesis Wnt signaling pathway, planar cell polarity pathway bone trabecula formation prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis regulation of branching involved in prostate gland morphogenesis regulation of establishment of planar polarity convergent extension involved in somitogenesis actin cytoskeleton organization substrate adhesion-dependent cell spreading convergent extension involved in axis elongation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-q2vkv:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline