Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Flotillin 2 Antibodies

          Invitrogen

          Flotillin 2 Polyclonal Antibody

          Advanced Verification
          1 Reference
          View all (18) Flotillin 2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Flotillin 2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (12)
          • Advanced Verification (2)
          Flotillin 2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Flotillin 2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 14

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Flotillin 2 Antibody (PA5-79268) in ICC/IF

          Immunocytochemistry analysis of Flotillin 2 using anti-Flotillin 2 antibody (Product # PA5-79268). Flotillin 2 was detected in a section of A431 cells. Enzyme antigen retrieval was performed using IHC enzyme antigen retrieval reagent for 15 mins. The cells were blocked with 10% goat serum and then incubated with 2μg/mL rabbit anti-Flotillin 2 antibody (Product # PA5-79268)overnight at 4°C. DyLight®550 Conju... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Flotillin 2 Antibody in Immunocytochemistry (ICC/IF)
          Flotillin 2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Flotillin 2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Flotillin 2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Flotillin 2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Flotillin 2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Flotillin 2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Flotillin 2 Antibody in Western Blot (WB)
          Flotillin 2 Antibody in Western Blot (WB)
          Flotillin 2 Antibody in Western Blot (WB)
          Flotillin 2 Antibody in Flow Cytometry (Flow)
          Flotillin 2 Antibody in Flow Cytometry (Flow)
          Flotillin 2 Antibody
          Flotillin 2 Antibody
          Flotillin 2 Polyclonal Antibody

          Product Details

          PA5-79268

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.5 µg/mL
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          0.5-1 µg/mL
          -

          Immunohistochemistry (Frozen) (IHC (F))

          0.5-1 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          2 µg/mL
          -

          Flow Cytometry (Flow)

          1-3 µg/1x10^6 cells
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E.coli-derived human Flotillin 2 recombinant protein (Position: K169-K344).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          500 µg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 5mg BSA

          Contains

          0.05mg sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2746384

          Product Specific Information

          Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.

          Positive Control - WB: human 293T whole cell, human Hela whole cell, human A549 whole cell, rat lung tissue, mouse lung tissue. IHC: human mammary cancer tissue, mouse intestine tissue, rat kidney tissue IHC-F: rat kidney tissue, rat small intestine tissue, mouse kidney tissue. ICC/IF: A431 cell. Flow: RH35 cell, U20S cell.

          Target Information

          Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. This gene encodes a caveolae-associated, integral membrane protein, which is thought to function in neuronal signaling.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Epidermal surface antigen; ESA; FLOT2; Flotillin 2 (epidermal surface antigen 1); Flotillin-2; Membrane component chromosome 17 surface marker 1; Membrane component chromosome 17 surface marker 1 homolog; membrane component, chromosome 17, surface marker 1 (35kD protein identified by monoclonal antibody ECS-1); OTTHUMP00000163513; REG-1; Reggie-1; reggie1-1

          View more View less

          Gene Aliases: AI573412; ECS-1; ECS1; ESA; ESA1; FLOT2; M17S1; Reg1; reggie-2

          View more View less

          UniProt ID: (Human) Q14254, (Mouse) Q60634, (Rat) Q9Z2S9

          View more View less

          Entrez Gene ID: (Human) 2319, (Mouse) 14252, (Rat) 83764

          View more View less

          Function(s)
          protease binding protein binding ionotropic glutamate receptor binding protein heterodimerization activity
          Process(es)
          membrane raft assembly cell adhesion negative regulation of gene expression protein localization to plasma membrane raft protein stabilization establishment of protein localization to plasma membrane negative regulation of amyloid precursor protein catabolic process positive regulation of establishment of T cell polarity epidermis development positive regulation of cell-cell adhesion positive regulation of I-kappaB kinase/NF-kappaB signaling regulation of myoblast differentiation protein localization to plasma membrane anterograde dendritic transport regulation of postsynaptic membrane neurotransmitter receptor levels
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-nnt95:80/100.66.79.76:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline