Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GSTA1/A2/A3/A4/A5 Antibodies

          Invitrogen

          GSTA1/A2/A3/A4/A5 Polyclonal Antibody

          1 Reference
          View all (2) GSTA1/A2/A3/A4/A5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite GSTA1/A2/A3/A4/A5 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          GSTA1/A2/A3/A4/A5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          GSTA1/A2/A3/A4/A5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GSTA1/A2/A3/A4/A5 Antibody (PA5-79335) in IHC (P)

          Immunohistochemical analysis of GSTA1/A2/A3/A4/A5 in paraffin-embedded section of human liver cancer tissues. Heat mediated antigen retrieval was performed in citrate buffer (pH6, epitope retrieval solution) for 20 mins. The tissue section was blocked with 10% goat serum. The tissue section was then incubated with 1μg/mL rabbit anti-GSTA1/A2/A3/A4/A5 antibody (Product # PA5-79335) overnight at 4°C. Biotinyl... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GSTA1/A2/A3/A4/A5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTA1/A2/A3/A4/A5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTA1/A2/A3/A4/A5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GSTA1/A2/A3/A4/A5 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          GSTA1/A2/A3/A4/A5 Polyclonal Antibody

          Product Details

          PA5-79335

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.5 µg/mL
          -

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          0.5-1 µg/mL
          -

          ELISA (ELISA)

          1-5 µg/mL (capture)
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E. coli-derived human GSTA1/A2/A3/A4/A5 recombinant protein (Position: A2-F222).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          500 µg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 5mg BSA

          Contains

          0.05mg sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2746451

          Product Specific Information

          Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.

          Positive Control - WB: human HepG2 whole cell, human HCCT tissue, human HCCP tissue, human HUH-7 whole cell, rat liver tissue, rat RH35 whole cell, mouse liver tissue. IHC: human lung cancer tissue, human liver cancer tissue, human liver cancer tissue.

          Target Information

          Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. These enzymes function in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione. The genes encoding these enzymes are known to be highly polymorphic. These genetic variations can change an individual's susceptibility to carcinogens and toxins as well as affect the toxicity and efficacy of some drugs. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-transferase belonging to the alpha class. The alpha class genes, located in a cluster mapped to chromosome 6, are the most abundantly expressed glutathione S-transferases in liver (hepatocytes) and kidney (proximal tubules). In addition to metabolizing bilirubin and certain anti-cancer drugs in the liver, the alpha class of these enzymes exhibit glutathione peroxidase activity, thereby protecting the cells from reactive oxygen species and the products of peroxidation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 13-hydroperoxyoctadecadienoate peroxidase; androst-5-ene-3,17-dione isomerase; detoxification enzyme; GHS transferase; glutathione S-alkyltransferase A1; glutathione S-alkyltransferase A2; glutathione S-alkyltransferase A3; glutathione S-alkyltransferase A4; glutathione S-aralkyltransferase A2; glutathione S-aralkyltransferase A3; glutathione S-aralkyltransferase A4; glutathione S-aryltransferase A1; glutathione S-aryltransferase A2; glutathione S-aryltransferase A3; glutathione S-aryltransferase A4; glutathione S-transferase; glutathione S-transferase (GST, EC 2.5.1.18); glutathione S-transferase 2; glutathione S-transferase 5.7; glutathione S-transferase A1; glutathione S-transferase A2; glutathione S-transferase A3; glutathione S-transferase A3 subunit; glutathione S-transferase A3-3; glutathione S-transferase A4; glutathione S-transferase A4-4; glutathione S-transferase A5; glutathione S-transferase A5-5; glutathione S-transferase GT41A; glutathione S-transferase Ha subunit 1; glutathione S-transferase Ha subunit 1 (EC 2.5.1.18); glutathione S-transferase Ha subunit 2 (EC 2.5.1.18); glutathione S-transferase Ya; glutathione S-transferase Ya subunit; glutathione S-transferase Ya-1; glutathione S-transferase Ya-2; glutathione S-transferase Ya1; glutathione S-transferase Ya3; glutathione S-transferase Yc; glutathione S-transferase Yc subunit; glutathione S-transferase Yc1 subunit; glutathione S-transferase Yc2 subunit; glutathione S-transferase Yk; glutathione transferase; glutathione transferase (EC 2.5.1.18); glutathione transferase A4-4; glutathione transferase YA subunit; Glutathione-S-transferase alpha type (Ya); glutathione-S-transferase, alpha type2; gluthione S-transferase subunit 1 (GST,EC 2.5.1.18); GST 1-1; GST 1a-1a; GST 1b-1b; GST 8-8; GST A1-1; GST A2-2; GST A4-4; GST A5-5; GST class-alpha member 1; GST class-alpha member 2; GST class-alpha member 3; GST class-alpha member 4; GST class-alpha member 5; GST HA subunit 1; GST HA subunit 2; GST K; GST Ya1; GST Ya2; GST Yc2; GST Yk; GST-gamma; GSTA1; GSTA4-4; ligandin; liver GST2; S-(hydroxyalkyl)glutathione lyase A1; S-(hydroxyalkyl)glutathione lyase A2; S-(hydroxyalkyl)glutathione lyase A3; S-(hydroxyalkyl)glutathione lyase A4; testicular tissue protein Li 80; testis tissue sperm-binding protein Li 59n; unnamed protein product

          View more View less

          Gene Aliases: Gm10639; GST 2-2; GST A3-3; GST AA; GST B; GST Yc1; GST-epsilon; GST2; Gst2-1; Gst2-2; Gst2-3; GST5.7; Gsta1; GSTA1-1; Gsta2; GSTA2-2; Gsta3; GSTA3-3; GSTA4-4; Gsta5; Gstc-2; Gstc2; GTA2; GTA3; GTA4; GTH1; GTH2; mGsta4; Yc1; Yc2

          View more View less

          UniProt ID: (Human) P08263, (Human) P09210, (Human) Q16772, (Human) O15217, (Human) Q7RTV2, (Rat) P04904, (Mouse) P13745, (Mouse) P10648, (Rat) P00502, (Mouse) P30115, (Rat) P46418, (Mouse) P24472, (Rat) P14942, (Rat) P04903

          View more View less

          Entrez Gene ID: (Human) 2938, (Human) 2939, (Human) 2940, (Human) 2941, (Human) 221357, (Rat) 24421, (Mouse) 14857, (Mouse) 14858, (Rat) 24422, (Mouse) 14859, (Rat) 494500, (Mouse) 14860, (Rat) 300850, (Rat) 494499, (Mouse) 100042314

          View more View less

          Function(s)
          glutathione transferase activity glutathione peroxidase activity steroid delta-isomerase activity fatty acid binding protein binding phospholipid-hydroperoxide glutathione peroxidase activity molecular carrier activity identical protein binding protein homodimerization activity dinitrosyl-iron complex binding glutathione binding toxic substance binding
          Process(es)
          glutathione metabolic process xenobiotic metabolic process response to bacterium epithelial cell differentiation response to stilbenoid xenobiotic catabolic process response to caloric restriction cellular oxidant detoxification ureteric bud development aflatoxin catabolic process prostaglandin metabolic process heme catabolic process linoleic acid metabolic process glutathione derivative biosynthetic process response to xenobiotic stimulus response to nutrient levels response to herbicide response to zinc ion response to nicotine cellular response to lithium ion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nv6df:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline