Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Laminin gamma-1,2,3 Antibodies

          Invitrogen

          Laminin gamma-1,2,3 Polyclonal Antibody

          Advanced Verification
          View all (2) Laminin gamma-1,2,3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Laminin gamma-1,2,3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (9)
          • Advanced Verification (1)
          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Laminin gamma-1,2,3 Antibody (PA5-79576) in IHC (P)

          Immunohistochemistry (Paraffin) analysis of Laminin gamma-1,2,3 in paraffin-embedded section of mouse skeletal muscle tissue using Laminin gamma-1,2,3 Polyclonal Antibody (Product # PA5-79576). Heat mediated antigen retrieval was performed in EDTA buffer (pH 8.0, epitope retrieval solution). The tissue section was blocked with 10% goat serum. The tissue section was then incubated with the primary antibody a... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Laminin gamma-1,2,3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Laminin gamma-1,2,3 Antibody in Western Blot (WB)
          Laminin gamma-1,2,3 Antibody in Western Blot (WB)
          Laminin gamma-1,2,3 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Laminin gamma-1,2,3 Polyclonal Antibody

          Product Details

          PA5-79576

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.5 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          0.5-1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Peptide mixture of laminin gamma1,2,3 (NKLNEIEGSLNKAKDEMKAS; DLEERVRRQRNHLHLLETSI; LQLDSHGALHHKLRQLEEES).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          500 µg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 4mg trehalose

          Contains

          0.05mg sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2746691

          Product Specific Information

          This antibody reacts will all 3 isoforms of Laminin gamma: Laminin gamma-1, gamma-2 and gamma-3.

          Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.

          Positive Control - WB: human A431 whole cell, human A549 whole cell, human PC-3 whole cell, human CACO-2 whole cell, rat NRK whole cell, rat RH35 whole cell, mouse NIH/3T3 whole cell, mouse HEPA1-6 whole cell. IHC: mouse heart tissue, mouse kidney tissue, rat cardiac muscle tissue, rat kidney tissue.

          Target Information

          Extracellular matrix (ECM) surrounds and separates most tissue compartments, determines tissue architecture and dynamically interacts with its surrounding cells. The basement membrane is a part of the ECM and is composed of collagens, proteoglycans, elastin and several glycoproteins of which laminin is the most abundant. Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains Dystroglycan is a major laminin receptor in muscle and epithelia, and nidogen is a high-affinity laminin-binding protein important for basement-membrane assembly.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: basement membrane protein; BM600-100kDa; cell-scattering factor 140 kDa subunit; CSF 140 kDa subunit; epiligrin subunit gamma; kalinin subunit gamma; kalinin/nicein/epiligrin 100 kDa subunit; ladsin 140 kDa subunit; LAMG3; lamimin, gamma 2; laminin B2 chain; laminin B2 precursor; laminin B2t chain; laminin gamma 3; laminin gamma2 chain; laminin subunit gamma-3-like; laminin, gamma 1; laminin, gamma 1 (formerly LAMB2); laminin, gamma 2; laminin, gamma 3; laminin-1 subunit gamma; laminin-10 subunit gamma; laminin-11 subunit gamma; laminin-12 subunit gamma; laminin-14 subunit gamma; laminin-15 subunit gamma; laminin-2 subunit gamma; laminin-3 subunit gamma; laminin-4 subunit gamma; laminin-5 subunit gamma; laminin-6 subunit gamma; laminin-7 subunit gamma; laminin-8 subunit gamma; laminin-9 subunit gamma; large adhesive scatter factor 140 kDa subunit; nicein subunit gamma; nicein, 100kD; nicein, 100kDa; S-LAM gamma; S-laminin subunit gamma; unnamed protein product

          View more View less

          Gene Aliases: 1110064A23Rik; B2e; B2T; BM600; CSF; EBR2; EBR2A; JEB3A; JEB3B; LAMB2; LAMB2T; LAMNB2; OCCM

          View more View less

          UniProt ID: (Human) P11047, (Human) Q13753, (Human) Q9Y6N6, (Mouse) Q9R0B6

          View more View less

          Entrez Gene ID: (Human) 3915, (Human) 3918, (Human) 10319, (Rat) 117036, (Mouse) 226519, (Rat) 192362, (Mouse) 16782, (Mouse) 23928, (Rat) 311862

          View more View less

          Function(s)
          extracellular matrix structural constituent extracellular matrix constituent conferring elasticity receptor agonist activity structural molecule activity glycosphingolipid binding protein binding heparin binding
          Process(es)
          axon guidance biological_process positive regulation of cell proliferation organ morphogenesis tissue development positive regulation of cell migration chromatin organization cell adhesion signal transduction gene expression cell migration extracellular matrix disassembly hair follicle morphogenesis neuron projection development hemidesmosome assembly substrate adhesion-dependent cell spreading hair cell differentiation tissue morphogenesis protein-containing complex assembly hair follicle cell proliferation positive regulation of skeletal muscle acetylcholine-gated channel clustering positive regulation of cell adhesion positive regulation of muscle cell differentiation regulation of basement membrane organization positive regulation of integrin-mediated signaling pathway endoderm development regulation of cell adhesion regulation of cell migration maintenance of blood-brain barrier regulation of embryonic development positive regulation of epithelial cell proliferation epidermis development cell morphogenesis sensory organ development visual perception astrocyte development retina development in camera-type eye
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-jvpnf:80/100.66.74.151:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline