Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: LQINQMFSVQ LSLGEQTWES EGSSIKKAQQ AVANKALTES TLPKPVQKPP KSNVNNNPGS ITPTVELNGL AMKRGEPAIY RPLDPKPFPN YRANYNFRGM YNQRYHCPVP KIFYVQLTVG NNEFFGEGKT RQAARHNAAM KALQ
Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 56%.
Staufen homolog 2 is a member of the family of double-stranded RNA (dsRNA)-binding proteins involved in the transport and/or localization of mRNAs to different subcellular compartments and/or organelles. These proteins are characterized by the presence of multiple dsRNA-binding domains which are required to bind RNAs having double-stranded secondary structures. Staufen homolog 2 shares 48. 5% and 59. 9% similarity with drosophila and human staufen, respectively. The exact function of Staufen homolog 2 is not known, but since it contains 3 copies of conserved dsRNA binding domain, it could be involved in double-stranded RNA binding events. Several transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: r-staufen protein; RNA binding protein; STAU2LL; RNA binding protein; STAU2LS; RNA binding protein; STAU2SL; RNA binding protein; STAU2SS; rStau; alternatively spliced; Stau2; staufen homolog 2; staufen, RNA binding protein, homolog 2; unnamed protein product
Gene Aliases: 39K2; 39K3
UniProt ID: (Human) Q9NUL3, (Rat) Q68SB1
Entrez Gene ID: (Human) 27067, (Rat) 171500
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support