Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: TTISLRQPPM AFQGISERQK ALRMV
Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 80%.
P2Y10 belongs to the family of G-protein coupled receptors, that are preferentially activated by adenosine and uridine nucleotides. Two alternatively spliced transcript variants encoding the same protein isoform have been found for this gene. P2Y10 has been reported to be expressed in lymphoid cells and in monocytes derived from dendritic cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G-protein coupled purinergic receptor P2Y10; lysophosphatidylserine receptor 2; p2Y purinoceptor 10; P2Y-like receptor; purinergic receptor P2Y, G-protein coupled, 10; purinergic receptor P2Y10; RP3-333E23.1; unnamed protein product
Gene Aliases: LPS2; LYPSR2; P2Y10
UniProt ID: (Human) O00398
Entrez Gene ID: (Human) 27334
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support