Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: EGHQSEGLRR FGGAFAPARC LTIAFKGRRK NLDLAAPTAE EAQRWVRGLT KLRARLDAMS QRERLDHWIH SYLHRADSNQ DSKMSFKEIK SLLRMVNVDM NDMYAYLLFK
Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 96%.
This gene encodes a member of the phospholipase C family, which catalyze the hydrolysis of phosphatidylinositol 4, 5-bisphosphate to generate the second messengers diacylglycerol and inositol 1, 4, 5-trisphosphate (IP3). Diacylglycerol and IP3 mediate a variety of cellular responses to extracellular stimuli by inducing protein kinase C and increasing cytosolic Ca(2+) concentrations. This enzyme localizes to the plasma membrane and requires calcium for activation. Its activity is inhibited by spermine, sphingosine, and several phospholipids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-3; phosphoinositide phospholipase C-delta-3; PLC delta3; unnamed protein product
Gene Aliases: PLC-delta-3
UniProt ID: (Human) Q8N3E9
Entrez Gene ID: (Human) 113026
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support