Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • H2AK5ac Antibodies

          Invitrogen

          H2AK5ac Polyclonal Antibody

          View all (8) H2AK5ac antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite H2AK5ac Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          H2AK5ac Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          H2AK5ac Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          H2AK5ac Antibody (PA5-85528) in ICC/IF

          Immunocytochemistry analysis of H2AK5ac in HeLa cells mock (left) and treated with 0.4 μM TSA for 18 hrs (right) fixed in 4% paraformaldehyde at RT for 15 min. Samples were incubated with H2AK5ac polyclonal antibody (Product # PA5-85528) using a dilution of 1:500 (green). Blue: Hoechst 33342 staining. Scale bar = 10 μm. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          H2AK5ac Antibody in Immunocytochemistry (ICC/IF)
          H2AK5ac Antibody in Western Blot (WB)
          H2AK5ac Antibody in Immunoprecipitation (IP)
          H2AK5ac Antibody in Dot Blot (DB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          H2AK5ac Polyclonal Antibody

          Product Details

          PA5-85528

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:20,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:1,000
          -

          Immunoprecipitation (IP)

          1:100-1:10,000
          -

          Dot blot (DB)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Carrier-protein conjugated synthetic peptide surrounding acetyl Lys5 of human Histone H2A. The exact sequence is proprietary.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7, with 1% BSA, 20% glycerol

          Contains

          0.01% thimerosal

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2792668

          Product Specific Information

          Keep as concentrated solution.

          Predicted reactivity: Rat (100%).

          Positive Control: HeLa.

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.

          ⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Acetyl-Histone H2; Acetyl-Histone H2A K5; Acetyl-Histone H2A Lys5; buforin I; H2A; H2A histone family member; H2A histone family member J; H2A histone family member V; H2A histone family member X; H2A histone family, member A; H2A histone family, member C; H2A histone family, member D; H2A histone family, member E; H2A histone family, member G; H2A histone family, member I; H2A histone family, member L; H2A histone family, member M; H2A histone family, member N; H2A histone family, member O; H2A histone family, member P; H2A histone family, member Q; H2A histone family, member R; H2A K5; H2A K5ac; H2A.1; H2A.F/Z; H2A.W histone; H2A/d; H2a/j; h2ad; H2ae; H2AFX; h2ai; H2ak; H2AK5; H2AK5ac; H2ALys5ac; H2AX histone; HIST1H3D; HIST2H2AA; HIST3H2A; histone 1, H2aa; histone 1, H2ab; histone 1, H2ac; histone 1, H2ad; histone 1, H2ae; histone 1, H2ag; histone 1, H2ai; histone 1, H2aj; histone 1, H2ak; histone 1, H2al; histone 1, H2am; histone 2, H2aa3; histone 2, H2aa4; histone 2, H2ab; histone 2, H2ac; Histone 2A; histone 3, H2a; histone cluster 1 H2A family member a; histone cluster 1 H2A family member b; histone cluster 1 H2A family member c; histone cluster 1 H2A family member d; histone cluster 1 H2A family member e; histone cluster 1 H2A family member g; histone cluster 1 H2A family member h; histone cluster 1 H2A family member i; histone cluster 1 H2A family member j; histone cluster 1 H2A family member k; histone cluster 1 H2A family member l; histone cluster 1 H2A family member m; histone cluster 1, H2aa; histone cluster 1, H2ab; histone cluster 1, H2ac; histone cluster 1, H2ad; histone cluster 1, H2ae; histone cluster 1, H2ag; histone cluster 1, H2ah; histone cluster 1, H2ai; histone cluster 1, H2ak; histone cluster 1, H2al; histone cluster 1, H2am; histone cluster 2 H2A family member a3; histone cluster 2 H2A family member a4; histone cluster 2 H2A family member b; histone cluster 2 H2A family member c; histone cluster 2, H2aa3; histone cluster 2, H2aa4; histone cluster 2, H2ab; histone cluster 2, H2ac; histone cluster 3 H2A; histone cluster 3, H2a; Histone H2A; histone H2A type 1-J; histone H2A-GL101; histone H2A.3; histone H2A.F/Z; histone H2A.x; histone H2A/a; histone H2A/g; histone H2A/l; histone H2A/m; histone H2A/p; histone H2A/ptl; histone H2A/q; histone H2A/r; histone H2A/s; histone H2AC; histone H2AD; histone H2AE; Histone H2AK5; Histone H2AK5ac; histone IIa; purine-rich binding element protein B; unnamed protein product

          View more View less

          Gene Aliases: bA317E16.2; dJ160A22.4; dJ193B12.1; dJ193B12.9; dJ221C16.4; dJ86C11.1; H2A; H2a-615; H2A-GL101; H2A.1; H2A.1b; H2A.2; H2A.3; H2A.i; H2A.X; H2A.Z-2; H2A/a; H2A/c; H2A/d; H2A/E; H2A/g; H2A/i; H2A/l; H2A/m; H2A/n; H2A/O; H2A/p; H2A/q; H2A/R; H2A/S; H2A/X; H2AA; H2AB; H2AFA; H2AFALii; H2AFC; H2AFD; H2AFE; H2AFG; H2AFI; H2AFJ; H2AFL; H2AFM; H2AFN; H2AFO; H2AFP; H2AFQ; H2AFR; H2AFV; H2AFX; H2AG; H2AH; H2AV; H2AW; HIST1H2AA; HIST1H2AB; HIST1H2AC; HIST1H2AD; HIST1H2AE; HIST1H2AG; HIST1H2AH; HIST1H2AI; HIST1H2AJ; HIST1H2AK; HIST1H2AL; HIST1H2AM; HIST2H2AA; HIST2H2AA3; HIST2H2AA4; HIST2H2AB; HIST2H2AC; HIST3H2A; HISTH2AA; pH2A/f; TH2A

          View more View less

          UniProt ID: (Human) Q96QV6, (Human) P0C0S8, (Human) Q96KK5, (Human) Q99878, (Human) Q6FI13, (Human) Q16777, (Human) Q8IUE6, (Human) Q7L7L0, (Human) P04908, (Human) Q93077, (Human) P20671, (Human) Q9BTM1, (Human) P16104, (Human) Q71UI9

          View more View less

          Entrez Gene ID: (Human) 221613, (Human) 8969, (Human) 85235, (Human) 8329, (Human) 8331, (Human) 8330, (Human) 8332, (Human) 8336, (Human) 8337, (Human) 723790, (Human) 8338, (Human) 317772, (Human) 92815, (Human) 8335, (Human) 8334, (Human) 3013, (Human) 3012, (Human) 55766, (Human) 3014, (Human) 94239

          View more View less

          Function(s)
          DNA binding structural constituent of chromatin protein heterodimerization activity protein binding enzyme binding damaged DNA binding histone binding chromatin-protein adaptor activity binding, bridging
          Process(es)
          chromatin organization negative regulation of cell proliferation heterochromatin assembly protein localization to CENP-A containing chromatin DNA damage checkpoint double-strand break repair nucleosome assembly cellular response to DNA damage stimulus response to ionizing radiation positive regulation of DNA repair protein localization to site of double-strand break nucleosome disassembly UV-damage excision repair
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-8lbpj:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0