Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PIK3CA Antibodies

          Invitrogen

          PIK3CA Polyclonal Antibody

          View all (36) PIK3CA antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PIK3CA Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          PIK3CA Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PIK3CA Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PIK3CA Antibody (PA5-87398) in ICC/IF

          Immunofluorescence analysis of PIK3CA in NIH/3T3 cells. Samples were incubated with PIK3CA Polyclonal antibody (Product # PA5-87398) using a dilution of 1:100 (40x lens). Blue: DAPI for nuclear staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PIK3CA Antibody in Immunocytochemistry (ICC/IF)
          PIK3CA Antibody in Immunocytochemistry (ICC/IF)
          PIK3CA Antibody in Immunocytochemistry (ICC/IF)
          PIK3CA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PIK3CA Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PIK3CA Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          PIK3CA Polyclonal Antibody

          Product Details

          PA5-87398

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000-1:20,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein (or fragment)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.64 mg/mL

          Amount

          64 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.09% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2804118

          Product Specific Information

          Positive Samples: HeLa

          Immunogen sequence: RLCLSICSVK GRKGAKEEHC PLAWGNINLF DYTDTLVSGK MALNLWPVPH GLEDLLNPIG VTGSNPNKET PCLELEFDWF SSVVKFPDMS VIEEHANWSV SREAGFSYSH AGLSNRLARD NELRENDKEQ LKAISTRDPL SEITEQEKDF LWSHRHYCVT IPEILPKLLL SVKWNSRDEV AQMYCLVKDW PPIKPEQAME

          Target Information

          Phosphatidylinositol 3-kinase is composed of an 85 kDa regulatory subunit and a 110 kDa catalytic subunit. The protein encoded by this gene represents the catalytic subunit, which uses ATP to phosphorylate PtdIns, PtdIns4P and PtdIns(4,5)P2. This gene has been found to be oncogenic and has been implicated in cervical cancers.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 2.7.1.153; EC:2.7.11.1; GRB1; lipid kinase; MGC142161; MGC142163; mutant phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha; p110alpha; phosphatidylinositol 3-kinase, catalytic, 110-KD, alpha; phosphatidylinositol 3-kinase, catalytic, alpha polypeptide; phosphatidylinositol 4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit alpha; phosphoinositide 3-kinase alpha; phosphoinositide-3-kinase catalytic alpha polypeptide; phosphoinositide-3-kinase, catalytic, alpha polypeptide; PI3-kinase p110 subunit alpha; PI3-kinase subunit alpha; PI3K alpha; PI3K-alpha; PI3Kalpha; pi3kca; PIK3R1; pk3ca; ptdIns-3-kinase subunit alpha; ptdIns-3-kinase subunit p110-alpha; serine/threonine protein kinase PIK3CA; unnamed protein product

          View more View less

          Gene Aliases: 6330412C24Rik; caPI3K; CCM4; CLAPO; CLOVE; CWS5; HMH; MCAP; MCM; MCMTC; p110; p110-alpha; p110alpha; PI3K; PI3K-alpha

          View more View less

          UniProt ID: (Human) P42336, (Mouse) P42337, (Rat) A0A0G2K344

          View more View less

          Entrez Gene ID: (Human) 5290, (Mouse) 18706, (Rat) 170911

          View more View less

          Function(s)
          protein serine/threonine kinase activity protein binding kinase activity 1-phosphatidylinositol-3-kinase activity protein kinase activator activity 1-phosphatidylinositol-4-phosphate 3-kinase activity insulin receptor substrate binding phosphatidylinositol-4,5-bisphosphate 3-kinase activity protein serine kinase activity
          Process(es)
          liver development regulation of protein phosphorylation glucose metabolic process insulin receptor signaling pathway response to hormone regulation of gene expression negative regulation of gene expression response to activity response to muscle inactivity cell migration actin cytoskeleton organization negative regulation of actin filament depolymerization cellular response to insulin stimulus response to muscle stretch phosphatidylinositol-3-phosphate biosynthetic process vascular endothelial growth factor signaling pathway TORC2 signaling regulation of multicellular organism growth response to leucine regulation of cellular respiration protein kinase B signaling negative regulation of neuron apoptotic process positive regulation of protein kinase activity phosphatidylinositol phosphorylation insulin-like growth factor receptor signaling pathway phosphatidylinositol-mediated signaling positive regulation of smooth muscle cell proliferation positive regulation of protein kinase B signaling relaxation of cardiac muscle adipose tissue development cellular response to glucose stimulus cellular response to hydrostatic pressure response to dexamethasone cardiac muscle cell contraction energy homeostasis regulation of actin filament organization autosome genomic imprinting response to butyrate positive regulation of protein localization to membrane negative regulation of fibroblast apoptotic process negative regulation of anoikis vasculature development epidermal growth factor receptor signaling pathway positive regulation of lamellipodium assembly negative regulation of macroautophagy platelet activation positive regulation of TOR signaling negative regulation of programmed cell death anoikis endothelial cell migration T cell receptor signaling pathway cardiac muscle contraction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-7d6nq:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline