Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • S-arrestin Antibodies

          Invitrogen

          S-arrestin Polyclonal Antibody

          View all (20) S-arrestin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite S-arrestin Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          S-arrestin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          S-arrestin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          S-arrestin Antibody (PA5-88939) in IHC (P)

          Immunohistochemistry analysis of S-arrestin in paraffin-embedded mouse retina. Samples were incubated with S-arrestin Polyclonal antibody (Product # PA5-88939) using a dilution of 1:200 (40x lens). Perform microwave antigen retrieval with 10 mM PBS buffer pH 7.2 before commencing with IHC staining protocol. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          S-arrestin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          S-arrestin Antibody in Western Blot (WB)
          S-arrestin Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          S-arrestin Polyclonal Antibody

          Product Details

          PA5-88939

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 1-405 of human SAG (NP_0005322)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.22 mg/mL

          Amount

          122 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.01% thimerosal

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2805237

          Product Specific Information

          Immunogen sequence: MAASGKTSKS EPNHVIFKKI SRDKSVTIYL GNRDYIDHVS QVQPVDGVVL VDPDLVKGKK VYVTLTCAFR YGQEDIDVIG LTFRRDLYFS RVQVYPPVGA ASTPTKLQES LLKKLGSNTY PFLLTFPDYL PCSVMLQPAP QDSGKSCGVD FEVKAFATDS TDAEEDKIPK KSSVRLLIRK VQHAPLEMGP QPRAEAAWQF FMSDKPLHLA VSLNKEIYFH GEPIPVTVTV TNNTEKTVKK IKAFVEQVAN VVLYSSDYYV KPVAMEEAQE KVPPNSTLTK TLTLLPLLAN NRERRGIALD GKIKHEDTNL ASSTIIKEGI DRTVLGILVS YQIKVKLTVS GFLGELTSSE VATEVPFRLM HPQPEDPAKE SYQDANLVFE EFARHNLKDA GEAEEGKRDK NDVDE; Positive Samples: Mouse eye, Rat eye

          Target Information

          S-arrestin, also known as retinal arrestin or visual arrestin, is part of a small gene family that includes four members. It primarily functions in the retina, specifically within rod and cone photoreceptors. S-arrestin plays a crucial role in phototransduction by participating in the inactivation of opsin, which is a key component in the visual signaling pathway. Upon light stimulation, activated opsin interacts with a G-protein-coupled receptor kinase (GRK), leading to its phosphorylation. S-arrestin then binds to phosphorylated opsin to terminate the G-protein signaling, hence contributing to the process of vision adaptation and regulation of neurotransmitter release at the photoreceptor-bipolar cell synapse. This interaction is vital for the proper functioning of the visual cycle, ensuring that 11-cis-retinal is regenerated through the retinoid cycle. Mutations in the S-arrestin gene, like other rhodopsin alleles, are linked to various retinal disorders. The gene's expression pattern is specific to the retina, underscoring its role in vision, and it is distinct from other members of the arrestin family like beta-arrestins which regulate G-protein-coupled receptors (GPCRs) outside of the retina.

          ⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 48 kDa protein; arrestin 1; arrestin-1; DKFZp686D1084; DKFZp686I1383; retina and pineal gland (arrestin); retinal S-antigen; rod arrestin; rod photoreceptor arrestin; S-AG; S-antigen; retina and pineal gland (arrestin); SAG; unnamed protein product; visual arrestin 1

          View more View less

          Gene Aliases: A930001K18Rik; Arr1; arrestin; Irbp; SAGMR; SANTI

          View more View less

          UniProt ID: (Mouse) P20443, (Rat) P15887

          View more View less

          Entrez Gene ID: (Mouse) 20215, (Rat) 25539

          View more View less

          Function(s)
          G-protein coupled receptor binding opsin binding spectrin binding phosphoprotein binding
          Process(es)
          G-protein coupled receptor internalization signal transduction sensory perception biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-swbc5:80/100.66.77.38:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0