Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TCIRG1 Antibodies

          Invitrogen

          TCIRG1 Polyclonal Antibody

          1 Published Figure
          1 Reference
          View all (10) TCIRG1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite TCIRG1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          • Published Figures (1)
          TCIRG1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TCIRG1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TCIRG1 Antibody (PA5-90425) in ICC/IF

          Immunofluorescence analysis of TCIRG1 in U-2 OS cells. Samples were incubated with TCIRG1 Polyclonal antibody (Product # PA5-90425) using a dilution of 1:100 (40x lens). Blue: DAPI for nuclear staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TCIRG1 Antibody in Immunocytochemistry (ICC/IF)
          TCIRG1 Antibody in Immunocytochemistry (ICC/IF)
          TCIRG1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TCIRG1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TCIRG1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TCIRG1 Antibody in Western Blot (WB)
          TCIRG1 Antibody in Western Blot (WB)
          TCIRG1 Polyclonal Antibody

          Product Details

          PA5-90425

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:5,000
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein (or fragment)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          2.2 mg/mL

          Amount

          220 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.05% BSA

          Contains

          0.09% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2806068

          Product Specific Information

          Immunogen sequence: MGSMFRSEEV ALVQLFLPTA AAYTCVSRLG ELGLVEFRDL NASVSAFQRR FVVDVRRCEE LEKTFTFLQE EVRRAGLVLP PPKGRLPAPP PRDLLRIQEE TERLAQELRD VRGNQQALRA QLHQLQLHAA; Positive Samples: U2OS, A-549, BxPC-3, U-937, HL-60, Mouse thymus, Mouse skeletal muscle, Rat thymus, Rat skeletal muscle; Cellular Location: Membrane, Multi-pass membrane protein

          Target Information

          The T-cell immune regulator 1 (TCIRG1) is the a3 subunit of the vacuolar-type proton transporting ATPase (V-ATPase), a proton pump in late endosomes and lysosomes that functions in the luminal acidification of these organelles. Genetic defects in the gene encoding this protein a responsible for a severe form of autosomal recessive osteopetrosis, a condition that leads to increased bone density, decreased bone strength, and inflammation in bone tissues. TCIRG1 is also required for the normal secretion of insulin and the bacteria-killing function of macrophages.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ATPase, H+ transporting, 116kD; ATPase, H+ transporting, lysosomal I; ATPase, H+ transporting, lysosomal V0 protein a; OC-116 kDa; osteoclastic proton pump; Osteoclastic proton pump 116 kDa subunit; osteosclerotic; specific 116-kDa vacuolar proton pump subunit; T cell immune response cDNA7 protein; T-cell immune regulator 1; T-cell immune response cDNA 7; T-cell immune response cDNA7 protein; T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein a; T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein A3; T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 subunit A3; TIRC7; transmembrane protein; V-ATPase 116 kDa subunit a 3; V-ATPase 116 kDa subunit a3; V-ATPase 116-kDa; V-ATPase a3; V-ATPase subunit a3; v-H+ATPase subunit a3; V-type proton ATPase 116 kDa subunit a; V-type proton ATPase 116 kDa subunit a 3; V-type proton ATPase 116 kDa subunit a isoform 3; V-type proton ATPase 116 kDa subunit a3; vacuolar proton translocating ATPase 116 kDa subunit A; Vacuolar proton translocating ATPase 116 kDa subunit a isoform 3

          View more View less

          Gene Aliases: a3; ATP6a3; Atp6i; ATP6N1C; ATP6V0A3; oc; OC-116; OC-116kDa; OC116; OPTB1; Stv1; TCIRG1; TIRC7; Vph1

          View more View less

          UniProt ID: (Human) Q13488

          View more View less

          Entrez Gene ID: (Human) 10312, (Rat) 293650, (Mouse) 27060

          View more View less

          Function(s)
          protein binding proton-transporting ATPase activity, rotational mechanism ATPase binding hydrogen ion transmembrane transporter activity ATP synthase
          Process(es)
          autophagosome assembly ossification osteoclast proliferation ion transport cellular calcium ion homeostasis autophagy apoptotic process inflammatory response cellular defense response vacuolar acidification protein catabolic process in the vacuole lysosomal lumen acidification positive regulation of cell proliferation regulation of proton transport response to silver ion gene expression regulation of gene expression immunoglobulin mediated immune response macroautophagy optic nerve development establishment of cell polarity myeloid cell differentiation B cell differentiation T cell differentiation osteoclast differentiation ruffle organization protein localization to organelle memory T cell activation T-helper 1 cell activation odontogenesis T cell homeostasis tooth eruption bone resorption regulation of osteoblast differentiation pH reduction homeostasis of number of cells regulation of insulin secretion intracellular pH reduction establishment of vesicle localization retina development in camera-type eye hematopoietic stem cell homeostasis enamel mineralization cellular response to cytokine stimulus phagosome acidification dentin mineralization hydrogen ion transmembrane transport ATP synthesis coupled proton transport ATP hydrolysis coupled proton transport proton transport vacuolar proton-transporting V-type ATPase complex assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-gkxk9:80/100.66.77.77:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline