Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MYOC Antibodies

          Invitrogen

          MYOC Polyclonal Antibody

          View all (13) MYOC antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite MYOC Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          MYOC Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MYOC Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MYOC Antibody (PA5-90794) in WB

          Western blot analysis of MYOC in extracts of various cell lines. Samples were incubated with MYOC Polyclonal antibody (Product # PA5-90794) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. Exposure time: 60s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MYOC Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          MYOC Polyclonal Antibody

          Product Details

          PA5-90794

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 245-504 of human MYOC (NP_0002521)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.46 mg/mL

          Amount

          46 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2806240

          Product Specific Information

          Immunogen sequence: CGELVWVGEP LTLRTAETIT GKYGVWMRDP KPTYPYTQET TWRIDTVGTD VRQVFEYDLI SQFMQGYPSK VHILPRPLES TGAVVYSGSL YFQGAESRTV IRYELNTETV KAEKEIPGAG YHGQFPYSWG GYTDIDLAVD EAGLWVIYST DEAKGAIVLS KLNPENLELE QTWETNIRKQ SVANAFIICG TLYTVSSYTS ADATVNFAYD TGTGISKTLT IPFKNRYKYS SMIDYNPLEK KLFAWDNLNM VTYDIKLSKM; Positive Samples: Raji, U-937, BT-474, Mouse brain; Cellular Location: Cell projection, Cytoplasmic vesicle, Endoplasmic reticulum, Golgi apparatus, Mitochondrion, Mitochondrion inner membrane, Mitochondrion intermembrane space, Mitochondrion outer membrane, Rough endoplasmic reticulum, cilium, exosome, extracellular matrix, extracellular space

          Target Information

          Myocilin is an extracellular protein expressed in the eye, including the retina, trabecular meshwork and ciliary body. Myocilin can form homomultimers in vivo and can also associate with components of the ECM via interactions with the Hep II domain of FibroPVRL1. In addition, myocilin interacts with myosin regulatory light chain, a component of the myosin motor protein complex. This interaction implies a role for myocilin in the actomyosin system, linking myocilin to the functional status of the trabecular meshwork (TM), which is responsible for controlling the intraocular pressure (IOP). Alterations in functions of the TM may lead to IOP elevation and development of glaucoma, a major cause of blindness. Myocilin is encoded by MYOC (also designated TIGR), a gene that maps to the GLC1A locus on chromosome 1q24. 3 and is susceptible to mutations. Mutations in the MYOC gene are specifically linked with primary open angle glaucoma (POAG), a blinding disease characterized by progressive loss of retinal ganglion cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: juvenile-onset open-angle glaucoma 1; mutated trabecular meshwork-induced glucocorticoid response protein; myocilin 55 kDa subunit; myocilin protein; myocilin trabecular meshwork inducible glucocorticoid response protein; trabecular meshwork inducible glucocorticoid response protein; trabecular meshwork-induced glucocorticoid response protein; unnamed protein product

          View more View less

          Gene Aliases: GLC1A; GPOA; JOAG; JOAG1; TIGR

          View more View less

          UniProt ID: (Human) Q99972, (Mouse) O70624

          View more View less

          Entrez Gene ID: (Human) 4653, (Mouse) 17926

          View more View less

          Function(s)
          fibronectin binding frizzled binding protein binding receptor tyrosine kinase binding myosin light chain binding
          Process(es)
          osteoblast differentiation negative regulation of cell-matrix adhesion signal transduction skeletal muscle hypertrophy myelination in peripheral nervous system positive regulation of cell migration neuron projection development negative regulation of Rho protein signal transduction non-canonical Wnt signaling pathway ERBB2-ERBB3 signaling pathway regulation of MAPK cascade clustering of voltage-gated sodium channels positive regulation of JNK cascade positive regulation of stress fiber assembly negative regulation of stress fiber assembly positive regulation of focal adhesion assembly positive regulation of protein kinase B signaling positive regulation of mitochondrial depolarization bone development positive regulation of substrate adhesion-dependent cell spreading
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-8lbpj:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0