Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TNFAIP3 Antibodies

          Invitrogen

          TNFAIP3 Polyclonal Antibody

          View all (29) TNFAIP3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite TNFAIP3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          TNFAIP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          TNFAIP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TNFAIP3 Antibody (PA5-96064) in IHC (P)

          Immunohistochemistry analysis of TNFAIP3 in paraffin-embedded human lung cancer. Samples were incubated with TNFAIP3 Polyclonal antibody (Product # PA5-96064) using a dilution of 1:50 (40x lens). Perform high pressure antigen retrieval with 10 mM citrate buffer pH 6.0 before commencing with IHC staining protocol. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TNFAIP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TNFAIP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TNFAIP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TNFAIP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TNFAIP3 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          TNFAIP3 Polyclonal Antibody

          Product Details

          PA5-96064

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:100-1:500
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 600-738 of human TNFAIP3 (NP_0062811)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.59 mg/mL

          Amount

          59 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2807866

          Product Specific Information

          Immunogen sequence: DRTGTSKCRK AGCVYFGTPE NKGFCTLCFI EYRENKHFAA ASGKVSPTAS RFQNTIPCLG RECGTLGSTM FEGYCQKCFI EAQNQRFHEA KRTEEQLRSS QRRDVPRTTQ STSRPKCARA SCKNILACRS EELCMECQH

          Positive Samples: Jurkat, HepG2, SW480, Mouse spleen; Cellular Location: Cytoplasm, Lysosome, Nucleus

          Target Information

          TNFAIP3, also known as A20, is located in chromosome band 6q23, a region that is often deleted in B cell lymphomas. Recently, it was identified as a tumor suppressor gene in Hodgkin lymphoma and several subtypes of non-Hodgkin lymphomas. TNFAIP3 was initially identified as a zinc-finger protein that is rapidly and transiently induced by TNF-a, inhibiting NF-kappa-B-dependent gene expression, and protecting cells from TNF-a-cytotoxicity. Overexpression of TNFAIP3 also inhibits the TLR2- and TLR4-mediated interleukin-8 synthesis in airway epithelial cells, suggesting that TNFAIP3 also acts as a negative regulator of TLR-mediated inflammatory responses, thereby protecting the host against harmful over-responses to pathogens. At least two isoforms of TNFAIP3 are known to exist.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: OTU domain-containing protein 7C; putative DNA-binding protein A20; RP11-356I2.3; TNF alpha-induced protein 3; Tnfaip3; TNFAIP3 (A20); tumor necrosis factor induced protein 3; tumor necrosis factor inducible protein A20; tumor necrosis factor, alpha induced protein 3; tumor necrosis factor, alpha-induced protein 3; unnamed protein product; zinc finger protein A20

          View more View less

          Gene Aliases: A20; AIFBL1; AISBL; OTUD7C; TNFA1P2; Tnfip3

          View more View less

          UniProt ID: (Human) P21580, (Mouse) Q60769

          View more View less

          Entrez Gene ID: (Human) 7128, (Rat) 683206, (Mouse) 21929

          View more View less

          Function(s)
          protease binding DNA binding catalytic activity ubiquitin-protein transferase activity thiol-dependent ubiquitin-specific protease activity protein binding zinc ion binding enzyme binding kinase binding identical protein binding ubiquitin binding Lys63-specific deubiquitinase activity
          Process(es)
          B-1 B cell homeostasis response to molecule of bacterial origin immune system process regulation of germinal center formation negative regulation of chronic inflammatory response protein deubiquitination negative regulation of protein ubiquitination response to muramyl dipeptide negative regulation of interleukin-1 beta production negative regulation of interleukin-2 production negative regulation of interleukin-6 production negative regulation of tumor necrosis factor production negative regulation of toll-like receptor 3 signaling pathway negative regulation of toll-like receptor 5 signaling pathway protein K11-linked deubiquitination negative regulation of viral-induced cytoplasmic pattern recognition receptor signaling pathway regulation of cell proliferation negative regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of protein catabolic process negative regulation of innate immune response negative regulation of smooth muscle cell proliferation negative regulation of inflammatory response negative regulation of B cell activation cellular response to hydrogen peroxide negative regulation of nucleotide-binding oligomerization domain containing 1 signaling pathway negative regulation of nucleotide-binding oligomerization domain containing 2 signaling pathway protein K63-linked deubiquitination protein K48-linked ubiquitination protein K48-linked deubiquitination cellular response to lipopolysaccharide protein deubiquitination involved in ubiquitin-dependent protein catabolic process tolerance induction to lipopolysaccharide establishment of protein localization to vacuole cellular response to oxygen-containing compound negative regulation of extrinsic apoptotic signaling pathway via death domain receptors positive regulation of hepatocyte proliferation negative regulation of CD40 signaling pathway negative regulation of endothelial cell apoptotic process negative regulation of extrinsic apoptotic signaling pathway regulation of tumor necrosis factor-mediated signaling pathway negative regulation of toll-like receptor 2 signaling pathway negative regulation of toll-like receptor 4 signaling pathway nucleotide-binding domain, leucine rich repeat containing receptor signaling pathway negative regulation of bone resorption regulation of defense response to virus by host regulation of vascular wound healing negative regulation of osteoclast proliferation marginal zone B cell differentiation negative regulation of granuloma formation regulation of immunoglobulin production response to wounding negative regulation of autophagy negative regulation of heterotypic cell-cell adhesion regulation of innate immune response
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-5rqsh:80/100.66.74.151:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0