Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD2AP Antibodies

          Invitrogen

          CD2AP Polyclonal Antibody

          View all (19) CD2AP antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD2AP Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          CD2AP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CD2AP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD2AP Antibody (PA5-96807) in ICC/IF

          Immunofluorescence analysis of CD2AP was performed using 70% confluent log phase A-431 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 1 hour at room temperature. The cells were labeled with CD2AP Polyclonal Antibody (Product # PA5-96807) at 1:100 dilution in 0.1% BSA, incubated at 4 degree celsius overnight and then with Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor Plus 488 (Product # A32731) at a dilu... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD2AP Antibody in Immunocytochemistry (ICC/IF)
          CD2AP Antibody in Western Blot (WB)
          CD2AP Antibody in Western Blot (WB)
          CD2AP Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          CD2AP Polyclonal Antibody

          Product Details

          PA5-96807

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunocytochemistry (ICC/IF)

          1:100
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 360-639 of human CD2AP (NP_0362521)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.57 mg/mL

          Amount

          57 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.01% thimerosal

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2808609

          Product Specific Information

          Positive Samples: A-431, Raji, MCF7, SW480, HL-60, Mouse kidney, Rat kidney; Cellular Location: Cell projection, Cytoplasm, cytoskeleton, ruffle

          Immunogen sequence: KYFSLKPEEK DEKSTLEQKP SKPAAPQVPP KKPTPPTKAS NLLRSSGTVY PKRPEKPVPP PPPIAKINGE VSSISSKFET EPVSKLKLDS EQLPLRPKSV DFDSLTVRTS KETDVVNFDD IASSENLLHL TANRPKMPGR RLPGRFNGGH SPTHSPEKIL KLPKEEDSAN LKPSELKKDT CYSPKPSVYL STPSSASKAN TTAFLTPLEI KAKVETDDVK KNSLDELRAQ IIELLCIVEA LKKDHGKELE KLRKDLEEEK TMRSNLEMEI EKLKKAVLSS

          Target Information

          This gene encodes a scaffolding molecule that regulates the actin cytoskeleton. The protein directly interacts with filamentous actin and a variety of cell membrane proteins through multiple actin binding sites, SH3 domains, and a proline-rich region containing binding sites for SH3 domains. The cytoplasmic protein localizes to membrane ruffles, lipid rafts, and the leading edges of cells. It is implicated in dynamic actin remodeling and membrane trafficking that occurs during receptor endocytosis and cytokinesis. Haploinsufficiency of this gene is implicated in susceptibility to glomerular disease.

          ⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: adapter protein CMS; cas ligand with multiple SH3 domains; Cas ligand with multiple Src homology (SH) 3 domains; Cas ligand with multiple Src homology 3 (SH3) domains; CD2AP; mesenchyme to epithelium transition protein, SH3 domains; mesenchyme-to-epithelium transition protein with SH3 domains 1; SH3 domain-containing adapter protein; SH3 domain-containing adaptor protein METS-1

          View more View less

          Gene Aliases: CMS; METS-1; Mets1

          View more View less

          UniProt ID: (Human) Q9Y5K6, (Mouse) Q9JLQ0, (Rat) F1LRS8

          View more View less

          Entrez Gene ID: (Human) 23607, (Mouse) 12488, (Rat) 316258

          View more View less

          Function(s)
          actin binding structural constituent of cytoskeleton protein binding SH3 domain binding clathrin binding protein binding, bridging phosphatidylinositol 3-kinase regulatory subunit binding identical protein binding cadherin binding actin filament binding vascular endothelial growth factor receptor binding beta-catenin binding macromolecular complex binding
          Process(es)
          immunological synapse formation kidney development liver development endothelium development protein phosphorylation lipid metabolic process apoptotic process inflammatory response response to oxidative stress actin filament organization cell proliferation male gonad development protein secretion response to wounding response to virus gene expression protein transport vesicle organization cell migration protein sumoylation actin filament polymerization cell differentiation protein catabolic process neuron projection development Rab protein signal transduction glomerulus development response to insulin negative regulation of transforming growth factor beta1 production regulation of actin cytoskeleton organization maintenance of blood-brain barrier neurotrophin signaling pathway nerve growth factor signaling pathway proteasome-mediated ubiquitin-dependent protein catabolic process protein kinase B signaling negative regulation of neuron apoptotic process cell-cell adhesion mediated by cadherin cell-cell junction organization filopodium assembly neurotrophin TRK receptor signaling pathway regulation of synaptic plasticity regulation of receptor-mediated endocytosis lymph node development collateral sprouting positive regulation of protein secretion synapse organization T cell receptor signaling pathway negative regulation of small GTPase mediated signal transduction protein heterooligomerization stress-activated MAPK cascade localization of cell adipose tissue development membrane organization ERK1 and ERK2 cascade response to transforming growth factor beta glomerular visceral epithelial cell differentiation renal albumin absorption cell-cell adhesion glucose import across plasma membrane positive regulation of protein localization to nucleus response to glial cell derived neurotrophic factor substrate-dependent cell migration, cell extension signal transduction protein-containing complex assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-q2vkv:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline