Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SHOX2 Antibodies

          Invitrogen

          SHOX2 Polyclonal Antibody

          View all (8) SHOX2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SHOX2 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          SHOX2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SHOX2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SHOX2 Antibody (PA5-96999) in WB

          Western blot analysis of SHOX2 in various lysates. Samples were incubated with SHOX2 Polyclonal antibody (Product # PA5-96999) using a dilution of 1:2,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. Exposure time: 60s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SHOX2 Antibody in Western Blot (WB)
          SHOX2 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          SHOX2 Polyclonal Antibody

          Product Details

          PA5-96999

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:5,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 96-355 of human SHOX2 (NP_0030213)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.01 mg/mL

          Amount

          101 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2808801

          Product Specific Information

          Immunogen sequence: ELDMGAAERS REPGSPRLTE GRRKPTKAEV QATLLLPGEA FRFLVSPELK DRKEDAKGME DEGQTKIKQR RSRTNFTLEQ LNELERLFDE THYPDAFMRE ELSQRLGLSE ARVQVWFQNR RAKCRKQENQ LHKGVLIGAA SQFEACRVAP YVNVGALRMP FQQDSHCNVT PLSFQVQAQL QLDSAVAHAH HHLHPHLAAH APYMMFPAPP FGLPLATLAA DSASAASVVA AAAAAKTTSK NSSIADLRLK AKKHAAALGL; Positive Samples: NCI-H460, Mouse skeletal muscle, Mouse liver; Cellular Location: Nucleus

          Target Information

          SHOX2 is a member of the homeo box family of genes that encode proteins containing a 60-amino acid residue motif that represents a DNA binding domain. Homeo box genes have been characterized extensively as transcriptional regulators involved in pattern formation in both invertebrate and vertebrate species. Several human genetic disorders are caused by aberrations in human homeo box genes. SHOX is a pseudoautosomal homeo box gene that is thought to be responsible for idiopathic short stature and implicated to play a role in the short stature phenotype of Turner syndrome patients. This gene is a member of the homeo box family of genes that encode proteins containing a 60-amino acid residue motif that represents a DNA binding domain. Homeo box genes have been characterized extensively as transcriptional regulators involved in pattern formation in both invertebrate and vertebrate species. Several human genetic disorders are caused by aberrations in human homeo box genes. SHOX is a pseudoautosomal homeo box gene that is thought to be responsible for idiopathic short stature and implicated to play a role in the short stature phenotype of Turner syndrome patients. This gene is considered to be a candidate gene for Cornelia de Lange syndrome. Alternative splicing has been observed at this locus and two variants, each encoding a distinct isoform, have been identified.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: homeobox protein Og12X; OG-12; OGI2X; paired family homeodomain protein Prx3; paired-related homeobox protein SHOT; Prx3A; Prx3A''; Prx3B; short stature homeobox 2; SHOX homologous gene on chromosome 3; unnamed protein product

          View more View less

          Gene Aliases: 6330543G17Rik; OG12; OG12X; Prx3; SHOT

          View more View less

          UniProt ID: (Human) O60902, (Rat) O35750, (Mouse) P70390

          View more View less

          Entrez Gene ID: (Human) 6474, (Rat) 25546, (Mouse) 20429

          View more View less

          Function(s)
          RNA polymerase II transcription factor activity, sequence-specific DNA binding DNA binding protein binding sequence-specific DNA binding sequence-specific double-stranded DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter skeletal system development regulation of heart rate sinoatrial node development sinoatrial valve development cardiac right atrium morphogenesis regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter nervous system development positive regulation of skeletal muscle fiber development positive regulation of axonogenesis muscle tissue morphogenesis cardiac pacemaker cell differentiation sinoatrial node cell development regulation of branching morphogenesis of a nerve osteoblast differentiation positive regulation of mesenchymal cell proliferation chondrocyte differentiation chondrocyte development heart valve development cardiac atrium morphogenesis embryonic limb morphogenesis regulation of chondrocyte differentiation embryonic forelimb morphogenesis positive regulation of smoothened signaling pathway positive regulation of transcription from RNA polymerase II promoter embryonic digestive tract morphogenesis embryonic morphogenesis skeletal system morphogenesis cartilage development embryonic skeletal joint morphogenesis cartilage development involved in endochondral bone morphogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-mwps6:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0