Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Caspase 1 p20 (Cleaved Asp296) Antibodies

          Invitrogen

          Caspase 1 p20 (Cleaved Asp296) Polyclonal Antibody

          10 Published Figures
          24 References

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Caspase 1 p20 (Cleaved Asp296) Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          • Published Figures (10)
          Caspase 1 p20 (Cleaved Asp296) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Caspase 1 p20 (Cleaved Asp296) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 13

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Caspase 1 p20 (Cleaved Asp296) Antibody (PA5-99390) in IHC (P)

          Immunohistochemistry analysis of Caspase 1 p20 (Cleaved Asp296) in rat heart tissue. The sample was formaldehyde fixed and a heat mediated antigen retrieval step in citrate buffer was performed. Samples were incubated with Caspase 1 p20 (Cleaved Asp296) polyclonal antibody (Product # PA5-99390) using a dilution of 1:100 (4°C overnight) followed by HRP conjugated anti-Rabbit secondary antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Caspase 1 p20 (Cleaved Asp296) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Caspase 1 p20 (Cleaved Asp296) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Caspase 1 p20 (Cleaved Asp296) Antibody in Western Blot (WB)
          Caspase 1 p20 (Cleaved Asp296) Antibody in Western Blot (WB)
          Caspase 1 p20 (Cleaved Asp296) Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Caspase 1 p20 (Cleaved Asp296) Antibody in Immunohistochemistry (IHC)
          Caspase 1 p20 (Cleaved Asp296) Antibody in Western Blot (WB)
          Caspase 1 p20 (Cleaved Asp296) Antibody in Immunocytochemistry (ICC/IF)
          Caspase 1 p20 (Cleaved Asp296) Antibody in Immunocytochemistry (ICC/IF)
          Caspase 1 p20 (Cleaved Asp296) Antibody in Immunohistochemistry (IHC)
          Caspase 1 p20 (Cleaved Asp296) Antibody in Western Blot (WB)
          Caspase 1 p20 (Cleaved Asp296) Antibody in Western Blot (WB)
          Caspase 1 p20 (Cleaved Asp296) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          PA5-99390

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 13 publications 13 publications

          Immunohistochemistry (IHC)

          -
          View 6 publications 6 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human CASP1(Accession P29466), corresponding to amino acid residues I280-V299.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2818323

          Product Specific Information

          Antibody detects endogenous levels of fragment of activated Caspase-1 resulting from cleavage adjacent to Asp296.

          Target Information

          Caspase-1 p20 (cleaved Asp296) refers to the large catalytic subunit generated during activation of pro-caspase-1, an inflammatory cysteine protease encoded by the CASP1 gene. Caspase-1 is synthesized as an inactive zymogen and becomes activated within multiprotein inflammasome complexes in response to infection, cellular stress, or danger-associated signals. Activation involves autoproteolytic cleavage at specific aspartic acid residues, including at Asp296, producing the p20 (large) and p10 (small) subunits that assemble into the active heterotetrameric enzyme. The p20 fragment contains part of the catalytic domain and is commonly used as a marker of caspase-1 activation. Active caspase-1 mediates the proteolytic maturation of pro-inflammatory cytokines such as interleukin-1β (IL-1β) and interleukin-18 (IL-18) and drives pyroptosis, an inflammatory form of programmed cell death. Detection of cleaved caspase-1 p20 (Asp296) is therefore widely used to assess inflammasome activation in immunology and inflammation research.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CASP-1; CASP1; caspase 1, apoptosis-related cysteine peptidase; Caspase-1 (Asp296 cleaved); Caspase-1 (cleaved Asp296); Cleaved Caspase-1 p20; Cleaved-Caspase-1; cysteine protease; IL-1 beta-converting enzyme; IL-1B converting enzyme; IL-1BC; IL1B-convertase; interleukin 1, beta, convertase; interleukin 1-B converting enzyme; Interleukin 1beta converting enzyme; interleukin-1 beta convertase; interleukin-1 beta-converting enzyme; p45; unnamed protein product

          View more View less

          Gene Aliases: ICE; IL1BC; P45

          View more View less

          UniProt ID: (Human) P29466, (Mouse) P29452

          View more View less

          Entrez Gene ID: (Human) 834, (Mouse) 12362, (Rat) 25166

          View more View less

          Function(s)
          endopeptidase activity cysteine-type endopeptidase activity protein binding cysteine-type peptidase activity cysteine-type endopeptidase activator activity involved in apoptotic process enzyme binding kinase binding cytokine binding identical protein binding CARD domain binding peptidase activity scaffold protein binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter response to hypoxia microglial cell activation plasma membrane repair positive regulation of cytokine production pattern recognition receptor signaling pathway proteolysis inflammatory response osmosensory signaling pathway myoblast fusion memory protein secretion response to xenobiotic stimulus response to bacterium regulation of autophagy programmed cell death protein processing protein autoprocessing protein catabolic process response to lipopolysaccharide positive regulation of interleukin-1 alpha production positive regulation of interleukin-1 beta production positive regulation of interleukin-18 production response to ATP interleukin-18-mediated signaling pathway defense response to bacterium regulation of apoptotic process positive regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of circadian sleep/wake cycle, non-REM sleep ceramide biosynthetic process regulation of inflammatory response positive regulation of inflammatory response defense response to Gram-negative bacterium protein maturation defense response to virus mitochondrial depolarization membrane hyperpolarization protein poly-ADP-ribosylation pyroptosis interleukin-1-mediated signaling pathway cellular response to lipopolysaccharide cellular response to interferon-gamma programmed necrotic cell death cytokine precursor processing signaling receptor ligand precursor processing AIM2 inflammasome complex assembly pyroptotic cell death positive regulation of NLRP3 inflammasome complex assembly response to paraquat positive regulation of tumor necrosis factor-mediated signaling pathway apoptotic process signal transduction positive regulation of neuron apoptotic process icosanoid biosynthetic process cellular response to mechanical stimulus positive regulation of release of cytochrome c from mitochondria midgut development response to toxic substance lung development animal organ development system development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0