Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • JNK1/JNK2/JNK3 Antibodies

          Invitrogen

          JNK1/JNK2/JNK3 Polyclonal Antibody

          View all (34) JNK1/JNK2/JNK3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite JNK1/JNK2/JNK3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (7)
          JNK1/JNK2/JNK3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          JNK1/JNK2/JNK3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          JNK1/JNK2/JNK3 Antibody (PA5-99528) in ICC/IF

          Immunocytochemistry analysis of JNK1/JNK2/JNK3 in HepG2 cells. The samples were fixed with PFA and permeabilized in 0.1% Triton X-100, then blocked in 10% serum for 45 minutes at 25°C. Samples were incubated with JNK1/JNK2/JNK3 polyclonal antibody (Product # PA5-99528) and mouse anti-beta tubulin antibody for 1 hour at 37°C. An AlexaFluor594 conjugated goat anti-rabbit IgG(H+L) (Red) and an AlexaFluor488 co... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          JNK1/JNK2/JNK3 Antibody in Immunocytochemistry (ICC/IF)
          JNK1/JNK2/JNK3 Antibody in Immunocytochemistry (ICC/IF)
          JNK1/JNK2/JNK3 Antibody in Immunocytochemistry (ICC/IF)
          JNK1/JNK2/JNK3 Antibody in Immunocytochemistry (ICC/IF)
          JNK1/JNK2/JNK3 Antibody in Western Blot (WB)
          JNK1/JNK2/JNK3 Antibody in Western Blot (WB)
          JNK1/JNK2/JNK3 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          JNK1/JNK2/JNK3 Polyclonal Antibody

          Product Details

          PA5-99528

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Pig, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human MAPK8(Accession P45983), corresponding to amino acid residues V158-L208.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2818461

          Product Specific Information

          Antibody detects endogenous levels of total JNK1/2/3.

          Target Information

          C-Jun N-terminal Kinase (JNK) is also known as Mitogen-activated protein kinase (MAPK8). It belongs to the MAPK superfamily of stress-activated protein kinases. MAPKs are Serine-threonine protein kinases that are activated in response to a variety of extracellular stimuli, and mediate signal transduction from the cell surface to the nucleus. JNK is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. JNK pathways are activated by stress and inflammatory signals. JNK is expressed as ten different isoforms due to differential mRNA splicing. The predominant forms are JNK1 and JNK2.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: c-Jun kinase 2; c-Jun N-terminal kinase 1; c-Jun N-terminal kinase 2; c-Jun N-terminal kinase 3; c-jun NH2-terminal kinase; FLJ12099; FLJ33785; janus kinase 1; janus kinase 2; JNK 46; JNK 55; JNK/SAPK alpha; JNK1 beta1 protein kinase; JNK3 alpha protein kinase; JNK3 beta1 protein kinase; JNK3 beta2 protein kinase; Jun kinase; JUN N-terminal kinase; MAP kinase 10; MAP kinase 8; MAP kinase 9; MAP kinase p49 3F12; MAPK 10; MAPK 8; MAPK 9; MAPK8; MAPK9; MGC50974; Mi; MK10; p54 gamma; p54-alpha; p54-beta; Phospho-JNK; protein kinase mitogen-activated 8; protein kinase, mitogen-activated 9; SAPK gamma; SAPK-alpha; SAPK-beta; SAPK/Erk/kinase 2; stress activated protein kinase alpha II; Stress activated protein kinase beta; stress-activated protein kinase 1; stress-activated protein kinase 1a; stress-activated protein kinase 1b; stress-activated protein kinase 1c; stress-activated protein kinase JNK1; stress-activated protein kinase JNK2; stress-activated protein kinase JNK3; unnamed protein product

          View more View less

          Gene Aliases: C230008H04Rik; JNK; JNK-46; JNK-55; JNK1; JNK1A2; JNK2; JNK21B1/2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; JNK3; JNK3A; JNK3B1; JNK3B2; p493F12; p54a; p54aSAPK; p54bSAPK; PRKM10; PRKM8; PRKM9; SAPb; SAPK; SAPK(beta); SAPK1; SAPK1a; SAPK1b; SAPK1c; SAPKC; Serk2

          View more View less

          UniProt ID: (Human) P53779, (Human) P45983, (Human) P45984, (Mouse) Q61831, (Rat) P49187, (Mouse) Q91Y86, (Rat) P49185, (Mouse) Q9WTU6, (Rat) P49186

          View more View less

          Entrez Gene ID: (Human) 5602, (Pig) 100521736, (Human) 5599, (Pig) 396610, (Pig) 396609, (Human) 5601, (Mouse) 26414, (Rat) 25272, (Mouse) 26419, (Rat) 116554, (Mouse) 26420, (Rat) 50658

          View more View less

          Function(s)
          protein kinase activity JUN kinase activity MAP kinase activity MAP kinase kinase activity protein binding ATP binding protein serine kinase activity protein serine/threonine kinase activity enzyme binding protein phosphatase binding histone deacetylase regulator activity histone deacetylase binding protein serine/threonine kinase binding protein serine/threonine/tyrosine kinase activity kinase activity kinesin binding mitogen-activated protein kinase kinase kinase binding non-receptor serine/threonine protein kinase
          Process(es)
          MAPK cascade protein phosphorylation signal transduction JNK cascade response to light stimulus Fc-epsilon receptor signaling pathway regulation of circadian rhythm cellular senescence response to oxidative stress JUN phosphorylation response to UV response to mechanical stimulus positive regulation of gene expression regulation of macroautophagy peptidyl-serine phosphorylation peptidyl-threonine phosphorylation positive regulation of cyclase activity positive regulation of cell killing negative regulation of protein binding regulation of protein localization cellular response to amino acid starvation cellular response to oxidative stress cellular response to reactive oxygen species positive regulation of apoptotic process negative regulation of apoptotic process positive regulation of protein metabolic process stress-activated MAPK cascade cellular response to lipopolysaccharide cellular response to mechanical stimulus energy homeostasis positive regulation of NLRP3 inflammasome complex assembly positive regulation of establishment of protein localization to mitochondrion positive regulation of macrophage derived foam cell differentiation positive regulation of protein ubiquitination positive regulation of proteasomal ubiquitin-dependent protein catabolic process modulation of synaptic transmission inflammatory response to wounding positive regulation of cytokine production involved in inflammatory response negative regulation of transcription from RNA polymerase II promoter locomotor rhythm neurotransmitter receptor transport to postsynaptic membrane vesicle-mediated transport in synapse cellular hyperosmotic response ossification neuron migration positive regulation of neuroblast proliferation double-strand break repair regulation of transcription, DNA-templated integrin-mediated signaling pathway neuromuscular junction development regulation of gene expression positive regulation of cardiac muscle cell apoptotic process positive regulation of cell migration positive regulation of microtubule polymerization neuron projection development positive regulation of glial cell apoptotic process ERBB signaling pathway response to hydrogen peroxide regulation of mRNA stability positive regulation of neuron apoptotic process NLRP3 inflammasome complex assembly regulation of neuron differentiation positive regulation of DNA replication regulation of centrosome cycle response to cadmium ion determination of dorsal identity dendrite morphogenesis SMAD protein signal transduction mRNA destabilization protein localization to tricellular tight junction protein poly-ADP-ribosylation cellular response to hydrogen peroxide cellular response to nitric oxide positive regulation of podosome assembly cellular response to oxygen-glucose deprivation type B pancreatic cell apoptotic process neuronal stem cell population maintenance programmed necrotic cell death regulation of dense core granule transport protein localization to site of double-strand break positive regulation of determination of dorsal identity positive regulation of neuron migration positive regulation of apoptotic signaling pathway glial cell apoptotic process response to steroid hormone neuron development cellular response to interleukin-1 cellular response to beta-amyloid release of cytochrome c from mitochondria positive regulation of protein phosphorylation positive regulation of prostaglandin biosynthetic process regulation of protein ubiquitination positive regulation of prostaglandin secretion positive regulation of chemokine production cellular response to UV positive regulation of cell differentiation positive regulation of transcription, DNA-templated regulation of JNK cascade extrinsic apoptotic signaling pathway response to xenobiotic stimulus response to toxic substance response to amine protein localization to mitochondrion cellular response to tumor necrosis factor cellular response to growth factor stimulus response to tetrachloromethane response to nerve growth factor
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nv6df:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline