Product Image
Thermo Scientific™
Personal DataRAM™ Accessories
Enhance functionality and performance using Personal DataRAM Accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
PDRZPZ-Pouch zeroing kit
PDRDCCDigital communication cable
PDRSSShoulder strap
PDRRARemote alarm interface unit
PDRACAC power supply and charger
PDRANCAnalog signal output cable
PDRCABelt clip kit
PDRZFZeroing filter kit
PDRCC1Soft shell carrying case
PDRUBUpper bumper kit
Catalog number PDRZP
Price (MXN)
-
Description:
Z-Pouch zeroing kit
Enhance functionality and performance using Thermo Scientific™ personal DataRAM Accessories. Available for conversion to passive sampling, from passive sampling or to active sampling.
For enhanced functionality and performance of both models pDR-1000AN and pDR1500. Conversion kits, routine parts and other supplies are available.
pDR1500 Standard Accessories:
  • Universal voltage power supply
  • 8 “AA” Batteries
  • Standard inlet
  • Zeroing tube
  • USB serial cable
  • Filter and filter holders (box of 100)
  • PC communications software disk
pDR-1000AN Standard Accessories:
  • Universal voltage power supply
  • Analog signal output cable
  • Zeroing kit
  • Belt clip kit
  • Carrying case
  • PC communications software disk
  • Digital communications cable
pDR1500 Optional Accessories:
  • Analog signal output cable
  • PM 10 PM 4 cyclone
  • PM 2.5 PM 1 cyclone
  • Carrying case
pDR-1000AN Optional Accessories:
  • Rechargeable (NiMH) battery pack
  • Remote alarm unit
  • Active sampling kit
  • Shoulder strap
  • Portable pump unit
  • Wall mounting bracketÂ
Attachable Pump Module pDR-PU:
  • Optional accessory designed for use with the personal DataRAM pDR-1000AN
  • A dual-chamber diaphram pump, a volumetric flow sensing and control unit, and a small rotameter-type flowmeter
  • Operates from either the optional rechargeable NiMH battery pack (pDR-BP) or from the AC line current using the power supply/charger (pDR-AC) supplied with the pDR-1000AN
  • Designed as a modular unit that can be used in various combinations
Rechargeable Battery Pack pDR-BP:
  • Rechargeable nickel-metal-hydride (NiMH) battery is designed to provide DC power
  • Allows operation for extended periods of time without requiring line power or battery replacement
Specifications
DescriptionZ-Pouch zeroing kit
For Use With (Equipment)D-1000 Series
Unit SizeEach