Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Proteins & Peptides ›
          • EGF Proteins
          Experience prolonged bioactivity in cell culture. Try Gibco Heat Stable Recombinant Proteins

          Gibco

          Human EGF Recombinant Protein

          5 References
          View all (14) EGF proteins

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support

          Cite Human EGF Recombinant Protein

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Testing Data (3)
          Human EGF Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.
          Human EGF Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Human EGF Protein (PHG0315) in Bioactivity

          Immunofluorescence analysis of signal transducer and activator of transcription 3 (STAT3) was performed using A549 cells treated with 100 ng/mL human EGF recombinant protein (Product # PHG0311) for 30 minutes. The cells were labeled with phospho-STAT3 (Ser727) recombinant rabbit monoclonal antibody (Product # MA5-33199) (Panel a & c; Green). The composite (b & d) represents the merged with DAPI (blue) and F-actin (red) staining. Cells treated with EGF displayed upregulated phospho-STAT3 (Ser727) in comparison with untreated cells. The imag... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Human EGF Protein in Bioactivity (Bioactivity)
          Human EGF Protein in Bioactivity (Bioactivity)
          Human EGF Protein in Bioactivity (Bioactivity)

          Product Details

          PHG0315

          Applications
          Tested Dilution
          Publications

          Bioactivity (Bioactivity)

          Assay-dependent
          View 5 publications 5 publications
          Product Specifications

          Species

          Human

          Published species

          Human

          Expression System

          E. coli

          Molecular weight

          6.2 kDa

          Class

          Recombinant

          Type

          Protein

          Purity

          >95% by SDS-PAGE

          Endotoxin concentration

          <0.1 ng/µg

          Activity

          ED50 < 0.4 ng/mL; determined by the dose-dependent proliferation of mouse BALB/3T3 cells.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Amount

          25 µg

          Purification

          purified

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Carrier-Free

          Reconstitution: Centrifuge the vial briefly before opening to bring the contents to the bottom. Reconstitute the lyophilized protein in sterile, PBS buffer to a concentration of 0.1-1.0 mg/mL. Make any further dilutions of the reconstituted protein in low endotoxin medium, or a buffered solution containing a carrier protein such as heat inactivated FCS or tissue culture grade BSA

          Storage: Store the lyophilized protein at 2°C to 8°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at ≤-20°C. Avoid repeated freeze-thaw cycles.

          Target Information

          EGF (Epidermal growth factor) exerts its actions by binding to the EGF Receptor, a 170 kDa protein kinase. Activation of EGFR initiates diverse cellular pathways in response to toxic environmental stimuli, or to EGF binding to the receptor, the EGFR forms homo- or heterodimers with other family members. Each dimeric receptor complex initiates a distinct signaling pathway by recruiting different Src homology 2 (SH2) containing effector proteins. EGF is far and wide expressed in kidney, cerebrum, prostrate and salivary glands. EGF acts as a potent mitogenic factor and the phosphorylated receptor recruits adapter proteins like GRB2 that activates complex downstream signaling cascades. EGF activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STAT modules. Research studies suggest the protein may also play important role in activating the NF-kappa-B signaling cascade. Defects in the EGGF gene are the cause of hypomagnesemia type 4 and dysregulation has been associated with the growth and progression of certain cancers.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: beta-urogastrone; Epidermal growth factor; H-EGF; unnamed protein product

          View more View less

          Gene Aliases: HOMG4; URG

          View more View less

          UniProt ID: (Human) P01133

          View more View less

          Entrez Gene ID: (Human) 1950

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch