Product Image
Invitrogen™

IBMX

IBMX
Have Questions?
Catalog NumberQuantity
PHZ11241 g
Catalog number PHZ1124
Price (USD)
477.00
Each
Quantity:
1 g
Price (USD)
477.00
Each

IBMX is a nonspecific inhibitor of phosphodiesterases that also possesses adenosine agonist activity.

Format/Formulation: Lyophilized

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Gene ID (Entrez)5595
Inhibitor PropertiesChemical
InhibitorsPhosphodiesterase Inhibitor
Molecular Weight (g/mol)222.2
Product TypeInhibitor
Purification MethodTLC
Quantity1 g
Shipping ConditionRoom Temperature
Specific Inhibit or TargetPhosphodiesterases (also has adenosine agonist activity)
SterilityNon-sterile
FormSolid
Research CategorySignal Transduction
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).

Frequently asked questions (FAQs)

Do you offer an alternative for the Revvity Health Sciences Inc IBMX Phosphodiesterase inhibitor (Cat. No. 50-211-8316) from Fisher Scientific?

Yes. We offer IBMX (Cat. No. PHZ1124), which can be found linked below:

IBMX (Cat. No. PHZ1124)

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.