IBMX - FAQs
IBMX - FAQs
View additional product information for IBMX - FAQs (PHZ1124)
1 product FAQs found
Do you offer an alternative for the Revvity Health Sciences Inc IBMX Phosphodiesterase inhibitor (Cat. No. 50-211-8316) from Fisher Scientific?
Yes. We offer IBMX (Cat. No. PHZ1124), which can be found linked below:
IBMX (Cat. No. PHZ1124)