Prionics™-Check WESTERN Kit (12000) - FAQs
Prionics™-Check WESTERN Kit (12000) - FAQs
View additional product information for Prionics™-Check WESTERN Kit (12000) - FAQs (PR12000)
1 product FAQs found
Do you offer the Antibody-AP from Prionics-Check WESTERN Kit (12000) as a stand-alone product?
The Antibody-AP is only available as part of the complete Prionics-Check WESTERN Kit (12000) (Cat. No. PR12000). It is not offered as a stand-alone product.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.