PowerEase™ Touch 120W Power Supply and XCell SureLock™ Mini-Cell and XCell II™ Blot Module Welcome Pack
Invitrogen™

PowerEase™ Touch 120W Power Supply and XCell SureLock™ Mini-Cell and XCell II™ Blot Module Welcome Pack

The PowerEase Touch 120W Power Supply* and XCell SureLock Mini-Cell and XCell II Blot Module Welcome Pack contains: • PowerEaseRead more
Have Questions?
Catalog NumberQuantity
PSC120SB1 pack
Catalog number PSC120SB
Price (USD)
1,339.00
Special offer
Online exclusive
Ends: 04-Dec-2026
2,060.00
Save 721.00
Each
Quantity:
1 pack
Price (USD)
1,339.00
Special offer
Online exclusive
Ends: 04-Dec-2026
2,060.00
Save 721.00
Each
The PowerEase Touch 120W Power Supply* and XCell SureLock Mini-Cell and XCell II Blot Module Welcome Pack contains:

• PowerEase Touch 120W Power Supply*
• PowerEase Touch 120W Power Supply (230 VAC)*
• PowerEase Touch 120W Power Supply (230 VAC) (Australia)*
• PowerEase Touch 120W Power Supply (230 VAC) (China)*
• XCell SureLock Mini-Cell and XCell II Blot Module

*dependent on region

About the PowerEase Touch 120W Power Supply
The Invitrogen PowerEase Touch 120W Power Supply is a medium-throughput power supply with a 4.3-inch backlit LCD touchscreen. The touchscreen and user interface design make setting up custom protocols or selecting one of the several pre-programmed gel electrophoresis and transfer methods a breeze. This power supply is ideal for DNA/RNA electrophoresis, SDS-PAGE, and native PAGE. The PowerEase Touch 120W Power Supply offers four sets of output jacks that can be used simultaneously and three modes: constant voltage, constant current, and constant power for flexibility of use and efficiency.

About the XCell SureLock Mini-Cell and XCell II Blot Module
The XCell SureLock Mini-Cell and XCell II Blot Module set is an electrophoresis and blotting system for mini gels. It includes the XCell SureLock Mini-Cell for running Invitrogen mini gels and the XCell II Blot Module for blotting up to two mini gels at a time. The XCell SureLock Mini-Cell holds gels firmly with a simple gel tension wedge. When the lever on the gel tension wedge is pushed forward into the locked position, an even, horizontal force is generated. This seals the gel/buffer core assembly firmly into position in the lower buffer chamber. The positive locking action of the gel tension wedge ensures a trouble-free, leak-free gel run every time.

The XCell II Blot Module allows you to easily transfer proteins or nucleic acids from mini gels to membranes. It fits into the XCell SureLock and XCell II Mini-Cells in place of the gel/buffer core assembly. It requires less than 200 mL of transfer buffer for western, southern, and northern transfers. Tough platinized titanium and stainless steel electrodes create a uniform electrical field without clamps or hinged gel holders. Maximum blot size is 9 cm x 9 cm.
Specifications
Ambient Temperature (English)Room Temperature
Dimensions (L x W x H)30.3 inch x 23 inch x 10.2 inch
Display4.3 inch backlit LCD
Max. Current500 mA
No. of Jacks4 (4 positive voltage and 4 negative voltage)
Quantity1 pack
Shipping ConditionRoom temperature
StackableYes
TimerUp to 999 min./step
WarrantyOne year
Wattage120 watts
Weight2.1 kg
For Use With (Application)DNA/RNA electrophoresis, SDS-PAGE, and native PAGE
Gel SizeMini
Max. Voltage300 VDC
Product LinePowerEase
Unit SizeEach