Fluorescein-PKC Substrate
Thermo Scientific™

Fluorescein-PKC Substrate

A fluorescein-labeled peptide substrate for the Protein Kinase C (PKC) family.
Have Questions?
Catalog NumberQuantity
PV35061 mg
Catalog number PV3506
Price (USD)
1,158.00
Each
Quantity:
1 mg
Price (USD)
1,158.00
Each

A fluorescein-labeled peptide substrate for the Protein Kinase C (PKC) family.

For use in combination with the LanthaScreen™ Tb-pSer (PKC Substrate) antibody (Cat. No. PV3560) to evaluate inhibitors in medium to high throughput screening applications.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Amino Acid SequenceFluorescein- RFARKGSLRQKNV
Assay EntryBiochemical terbium phospho-specific activity
Label or DyeFITC (Fluorescein)
Molecular Weight (g/mol)1919 Da
Quantity1 mg
Shipping ConditionDry Ice
SubstrateFluorescein-PKC
Target EntryMAP4K5, PRKCH, PRKCQ, PRKX, ROCK2, PRKCD, NEK2, PRKCG, SGK, MAP4K2, MINK1, SGK2, PRKCA, PRKCI, AURKB, TAOK2, MAP4K4, MARK4, DYRK4, PHKG1, PRKCZ, ROCK1, RPS6KA1, PRKCB1, STK24, PRKCB2, RPS6KA2, SGKL, STK25, PRKCE, MST4
Validated KinasesAURKB, DYRK4, MARK4, MAP4K2, MAP4K4, MAP4K5, MINK1, MST4, NEK2, PHKG1, PKC, PRKX, ROCK2, RPS6KA1, RPS6KA2, STK24, STK25, SGK, SGK2, SGKL, TAOK2, TBK1
Detection MethodFluorescence
Substrate PropertiesPeptide-Based Substrate
Substrate TypeKinase Substrate
Target EnzymeSerine/Threonine Kinases
Unit SizeEach
Contents & Storage
1 mg/mL Fluorescein PKC peptide substrates in water. Store at -20°C and protect from light.