Product Image
Thermo Scientific™

PI Lipid Kinase Substrate

Specifically optimized for use with the Class II (PIK3C2A & PIK3C2B) PI3 kinases.
Have Questions?
Catalog NumberQuantity
PV5371800 nmol
Catalog number PV5371
Price (USD)
1,164.00
Each
Quantity:
800 nmol
Price (USD)
1,164.00
Each

Specifically optimized for use with the Class II (PIK3C2A & PIK3C2B) PI3 kinases. These vesicles consist of 100% phosphatidylinositol (PI).

For use with the Adapta™ Universal Kinase Assay (Cat. No. PV5099).

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Assay EntryBiochemical ADP accumulation
Quantity800 nmol
Shipping ConditionDry Ice
SubstratePhosphatidylinositol (PI) vesicles
Target EntryPIK3C2B, PIK3C2A
Validated KinasesPIK3C2B
Detection MethodFluorescence
FormFrozen
Substrate PropertiesLipid-Based Substrate, Peptide-Based Substrate
Substrate TypeKinase Substrate
Target EnzymeLipid Kinases
Unit SizeEach
Contents & Storage
PI Lipid Kinase Substrate in 50 mM HEPES, pH 7.5

Store at -20°C.