Kinase Tracer 314, 25 μM

Kinase Tracer 314, 25 μM

In the LanthaScreen™ Eu Kinase Binding Assay, binding of an Alexa Fluor™ conjugate or “tracer” such as Kinase Tracer 314Read more
Have Questions?
Catalog NumberQuantity
PV608725 μL
Catalog number PV6087
Price (USD)
662.00
Each
Quantity:
25 μL
Price (USD)
662.00
Each
In the LanthaScreen™ Eu Kinase Binding Assay, binding of an Alexa Fluor™ conjugate or “tracer” such as Kinase Tracer 314 to a kinase is detected by addition of a Eu-labeled anti-tag antibody. Binding of the tracer & antibody to a kinase results in a high degree of FRET, whereas displacement of the tracer with a kinase inhibitor results in a loss of FRET. Unlike many kinase activity assays, this assay is a simple mix and read assay, with no development steps. Learn More and See Which Kinases Have Been Validated Using This Assay
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayKinase Binding Assay
Assay EntryBiochemical binding/kinetic
Excitation/Emission340/615, 665
For Use With (Application)TR-FRET
For Use With (Equipment)Microplate Reader
Format384-well Plate
Label or DyeAlexa Fluor™ 647
Quantity25 μL
ReadoutKinetic
Shipping ConditionDry Ice
Target EntryPIK3CG, PIK3CD/PIK3R1, mTOR, FRAP1
Validated KinasesPIK3CD, PIK3CG, FRAP1 (mTOR)
TypeKinase Tracer
Unit SizeEach
Contents & Storage
Store at -20°C.