Kinase Tracer 1710, 25 μM

Kinase Tracer 1710, 25 μM

In the LanthaScreen™ Eu Kinase Binding Assay, binding of an Alexa Fluor™ conjugate or “tracer” such as Kinase Tracer 1710Read more
Have Questions?
Catalog NumberQuantity
PV608825 μL
Catalog number PV6088
Price (USD)
666.00
Each
Quantity:
25 μL
Price (USD)
666.00
Each
In the LanthaScreen™ Eu Kinase Binding Assay, binding of an Alexa Fluor™ conjugate or “tracer” such as Kinase Tracer 1710 to a kinase is detected by addition of a Eu-labeled anti-tag antibody. Binding of the tracer & antibody to a kinase results in a high degree of FRET, whereas displacement of the tracer with a kinase inhibitor results in a loss of FRET. Unlike many kinase activity assays, this assay is a simple mix and read assay, with no development steps. Learn More and See Which Kinases Have Been Validated Using This Assay
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayKinase Binding Assay
Assay EntryBiochemical binding/kinetic
Excitation/Emission340/615, 665
For Use With (Application)TR-FRET
For Use With (Equipment)Microplate Reader
Format384-well Plate
Label or DyeAlexa Fluor™ 647
Quantity25 μL
ReadoutKinetic
Shipping ConditionDry Ice
Target EntryABL1 G250E, ABL1 E255K, ABL1 Y253F, ABL1, PIK3CA/PIK3R1, WNK3
Validated KinasesPIK3CA, ABL1, ABL1 E255K, ABL1 G250E, ABL1 Y253F, WNK3
TypeKinase Tracer
Unit SizeEach
Contents & Storage
Store at -20°C.