Search
View additional product information for Qubit™ microRNA Assay Kits - FAQs (Q32880, Q32881)
4 product FAQs found
The exact excitation/emission wavelength information is proprietary. Here are the approximate excitation/emission wavelengths:
- Qubit dsDNA HS Assay: ~500 nm/ ~530 nm
- Qubit dsDNA BR Assay: ~510 nm/ ~530 nm
- Qubit ssDNA Assay: ~490 nm/ ~520 nm
- Qubit RNA HS Assay: ~640 nm/ ~670 nm
- Qubit RNA BR Assay: ~640 nm/ ~670 nm
- Qubit microRNA Assay: ~500 nm/ ~520 nm
- Qubit Protein Assay: ~470 nm/ ~570 nm
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.
If your sample may contain both DNA and RNA, one may use either (or both) the DNA and RNA Qubit kits and compare with samples treated with either RNase or DNase to get an accurate determination of DNA or RNA, respectively.