Quant-iT™ microRNA Assay Kit - FAQs
Quant-iT™ microRNA Assay Kit - FAQs
View additional product information for Quant-iT™ microRNA Assay Kit - FAQs (Q32882)
1 product FAQs found
Why am I getting negative fluorescence values with my Qubit Assays?
Negative fluorescence is a physical impossibility. It is an artifact from software autocorrecting for background signal. This means your reader is picking up and subtracting out background light at the cost of your data. Make sure to do a buffer-only control and assess the type of signal. You may need to switch to a different plate.