Quant-iT™ RNA HS Reagent, 1 mL - FAQs
Quant-iT™ RNA HS Reagent, 1 mL - FAQs
View additional product information for Quant-iT™ RNA Assay Kits and Quant-iT RNA HS Reagent - FAQs (Q33140, Q10213, Q32884, Q33225)
1 product FAQs found
Why am I getting negative fluorescence values with my Qubit Assays?
Negative fluorescence is a physical impossibility. It is an artifact from software autocorrecting for background signal. This means your reader is picking up and subtracting out background light at the cost of your data. Make sure to do a buffer-only control and assess the type of signal. You may need to switch to a different plate.