Qubit™ Flex Pyrogen-Free Tube Strips - FAQs
Qubit™ Flex Pyrogen-Free Tube Strips - FAQs
View additional product information for Qubit™ Flex Pyrogen-Free Tube Strips - FAQs (Q32893)
1 product FAQs found
Are Qubit Flex Pyrogen-Free Tube Strips compatible with other Qubit assays besides the Qubit Endotoxin Detection Assay?
Qubit Flex Pyrogen-Free Tube Strips have not been tested with other Qubit assays. We recommend using Qubit Flex Assay Tube Strips (Cat. No. Q33252) for dsDNA, ssDNA, RNA, miRNA, and protein quantification.