Can the Qubit RNA IQ Assay Kits (Cat. No. Q33221, Q33222) be run on a plate reader for high throughput screening of RNA integrity?
Qubit RNA IQ Assay Kits are not compatible with plate readers. The IQ value calculations are performed by the Qubit instrument, which involves proprietary algorithms that cannot be replicated on other devices. For accurate results, use the Qubit instrument as recommended.