What is the expiration date for the Quant-iT 1X dsDNA Assay Kits, high sensitivity (HS) and broad range (BR) (Cat. Nos. Q33232, Q3367)?
The expiration date for the Quant-iT 1X dsDNA Assay Kits, high sensitivity (HS) and broad range (BR) (Cat. Nos. Q33232, Q3367) is 6 months from the date of receipt.