QuantiGene™ Singleplex Assay Kit, 50 plates - FAQs
QuantiGene™ Singleplex Assay Kit, 50 plates - FAQs
View additional product information for QuantiGene™ Singleplex Assay Kit - FAQs (QS0012, QS0013, QS0016)
1 product FAQs found
How does the QuantiGene Singleplex Assay Kit (Cat. No. QS0011) differ from the new kit (Cat. Nos. QS0012, QS0013, QS0014, QS0016)?
The QuantiGene Singleplex Assay Kit has been reformulated such that the Pre-Amplifier Solution and Amplifier Solution now come pre-diluted, which streamlines the workflow, and also these solutions can be shipped and stored at 2-8 degrees C now instead of -20 degrees C. The Blocking Reagent is also now shipped and stored at 2-8 degrees C. Hence, the QuantiGene Singleplex Assay Kit (Cat. No. QS0011) has been discontinued and replaced with the new kit (Cat. Nos. QS0012, QS0013, QS0014, QS0016).