ViewRNA™ Colorimetric Tissue Core Kit and Individual Modules
Invitrogen™
ViewRNA™ Colorimetric Tissue Core Kit and Individual Modules
The ViewRNA Tissue Core Kit and Modules enable sensitive and specific colorimetric detection for RNA in situ hybridization (ISH) in tissue using branched DNA (bDNA) signal amplification technology and the Fast Red, Fast Blue, and DAB substrates. ViewRNA Tissue probe sets are sold separately.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
QVT0430ViewRNA DAB Module
QVT0410ViewRNA Fast Red Module
QVT0420ViewRNA Fast Blue Module
QVT0400ViewRNA Tissue Core Kit
QVT0400AViewRNA Tissue Core Reagents
QVT0400BViewRNA Tissue Wash Buffers
Catalog number QVT0430
Price (MXN)
15,679.52
Each
Product Type:
ViewRNA DAB Module
Price (MXN)
15,679.52
Each
ViewRNA Colorimetric Tissue assays are RNA in situ hybridization (ISH) assays for the reliable detection of mRNA targets within tissue sections at single-copy sensitivity. The ViewRNA assays use proprietary chemistry for the target-specific probe sets along with branched DNA (bDNA) technology to amplify target-specific signal and achieve superior signal-to-noise ratios compared to traditional colorimetric ISH signal amplification methods, which often amplify both the signal and background.
When combined with a ViewRNA Tissue probe set (purchased separately), the ViewRNA Tissue Core Kit and ViewRNA Modules enable the colorimetric detection of mRNA targets in tissue sections using the Fast Red, Fast Blue, and DAB substrates.
ViewRNA Tissue assays are RNA in situ hybridization (ISH) assays for the reliable detection of one or two mRNA targets within tissue sections. Visualize any gene in any tissue with single-copy sensitivity and no radioactivity. And in case your needs are not met by one of our 6500 catalog probe sets, we have decades of bioinformatics expertise and will design custom probe sets to any mRNA target of interest, free of charge.
With ViewRNA Tissue assays, you can achieve:
• Robust, single mRNA molecule detection in tissue sections
• Accuracy and specificity inherent to branched DNA amplification technologies
• Simultaneous detection of two RNA targets
The ViewRNA Tissue Assay is compatible with formalin-fixed, paraffin-embedded (FFPE) and cryopreserved tissue. Whereas traditional ISH technologies often amplify both signal and background, ViewRNA assays use branched DNA (bDNA) technology to amplify target-specific signal and achieve superior signal-to-noise ratios.
To meet various experimental needs, the ViewRNA Tissue assay is provided in a variety of kits and modules. The ViewRNA Tissue Core Kit and ViewRNA Modules can be purchased separately in different combinations to mix and match and create a colorimetric ViewRNA assay for detection of up to two RNA targets. For added convenience, ViewRNA 1-Plex and 2-Plex Kits containing the ViewRNA Tissue Core Kit and ViewRNA Modules can also be purchased, then supplemented with the ViewRNA Tissue Core Kit and ViewRNA Modules as needed for additional assays or substrates. 
ViewRNA Tissue Core Kit
The ViewRNA Tissue Core Kit includes the ViewRNA Tissue Core Reagents (2-8°C reagents) and ViewRNA Tissue Wash Buffers (room temperature buffers) containing the reagents for in situ hybridization to RNA targets, but does not contain the specific target probes or colorimetric substrate for detection. The ViewRNA Tissue Core Kit can be combined with the ViewRNA Fast Red, Fast Blue, and DAB Modules which provide the substrates for RNA detection in ViewRNA colorimetric ISH tissue staining. The ViewRNA Tissue Core Reagents and ViewRNA Tissue Wash Buffers can also be purchased separately. Sufficient reagents are provided to stain 24 tissue sections.
ViewRNA Fast Red Module
The ViewRNA Fast Red Module includes the alkaline phosphatase (AP) substrate Fast Red to detect a single mRNA target with red colored precipitate for ViewRNA colorimetric ISH tissue staining. The Fast Red substrate is for use with ViewRNA Type 1 or Type 6 Probe Sets (ViewRNA probe sets are sold separately). Sufficient reagents are provided to stain 24 tissue sections.
ViewRNA Fast Blue Module
The ViewRNA Fast Blue Module includes the alkaline phosphatase (AP) substrate Fast Blue to detect a single mRNA target with blue colored precipitate for ViewRNA colorimetric ISH tissue staining. The Fast Blue substrate is for use with ViewRNA Type 6 or Type 1 Probe Sets (ViewRNA probe sets are sold separately). Sufficient reagents are provided to stain 24 tissue sections.
ViewRNA DAB Module
The ViewRNA DAB Module includes the horseradish peroxidase (HRP) substrate DAB to detect a single mRNA target with brown colored staining for ViewRNA colorimetric ISH tissue staining. The DAB substrate is for use with ViewRNA Type 4 Probe Sets (ViewRNA probe sets are sold separately). Sufficient reagents are provided to stain 24 sections.
For Research Use Only.
Specifications
For Use With (Application)In Situ Hybridization (ISH)
Product LineViewRNA
Product TypeViewRNA DAB Module
Quantity24 Assays
Shipping ConditionWet ice
Detection MethodChromogenic
Final Product TypeRNA (Labeled)
FormatKit
Labeling TargetRNA (General)
Unit SizeEach
Contents & Storage
• 720 μL DAB Solution (2–8°C)
• 24 mL DAB Reaction Buffer (2–8°C)
• 20 mL Peroxidase Quencher (2–8°C)
• 45 μL Label Probe 4 - HRP (2–8°C, do not freeze)
Frequently asked questions (FAQs)
Do you offer an alternative for the discontinued ViewRNA ISH Tissue Assay kits (2-plex) (Cat. No. QVT0012 and QVT0013)?
Currently, we offer the ViewRNA Colorimetric Tissue Assays in three formats:
- ViewRNA Colorimetric Tissue Core Kit (Cat. No. QVT0400) and  Individual modules for Fast Red (Cat. No. QVT0410), Fast Blue (Cat. No. QVT0420) and DAB (Cat. No. QVT0430) signal development
- ViewRNA Colorimetric Tissue 1-Plex Kits (Cat. No. QVT0420C, QVT4420C, QVT0410C, QVT4410C, QVT0430C, QVT4430C)
- ViewRNA Colorimetric Tissue 2-Plex Kits (Cat. No. QVT0433C, QVT4433C, QVT0412C, QVT4412C, QVT0422C, QVT4422C)
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Do you have an alternative for the ViewRNA ISH Tissue Assay Kit (2-plex) and ViewRNA Chromogenic Signal Amplification (1-plex) Kits (Cat. Nos. QVT0012, QT0200)?
Unfortunately, there is no exact comparable kit to the discontinued kit ViewRNA Chromogenic Signal Amplification Kit (1-plex) (Cat. No. QVT0200) as the use of the Naphthol Buffer is no longer required for signal amplification.
The closest replacement we currently offer is the ViewRNA Colorimetric Tissue Core Kit and Individual Modules:
ViewRNA Colorimetric Tissue Core Kit and Individual Modules (Cat. No. QVT0400)
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.