Product Image
Invitrogen™
ViewRNA™ Fast Red Buffer and Substrate Kit
ViewRNA Fast Red Buffer and Substrate Kit is an individual component of the ViewRNA Chromogenic Signal Amplification Kit.
Have Questions?
Catalog NumberQuantity
QVT050424 assays
Catalog number QVT0504
Price (USD)
844.00
Each
Quantity:
24 assays
Price (USD)
844.00
Each
ViewRNA Fast Red Buffer and Substrate Kit is an individual component of the ViewRNA Chromogenic Signal Amplification Kit, 24 assays (Cat. No. QVT0200). It is a red substrate used for the detection of alkaline phosphatase activity.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeRNA In Situ Hybridization Buffer and Substrate Kit
Quantity24 assays
FormatKit
Unit SizeEach