Product Image
Invitrogen™
Fast Blue Substrate (20 mL Blue Buffer + 420 uL each Blue Reagents 1-3)
Fast Blue Substrate (20 mL Blue Buffer + 420 uL each Blue Reagents 1-3) is an individual component of the ViewRNA™ ISH Tissue Assay Kits, 2-Plex
Have Questions?
Catalog NumberQuantity
QVT050624 Assays
Catalog number QVT0506
Price (USD)
854.00
Each
Quantity:
24 Assays
Price (USD)
854.00
Each
Fast Blue Substrate (20 mL Blue Buffer + 420 uL each Blue Reagents 1-3) is an individual component of the ViewRNA™ ISH Tissue Assay Kits, 2-Plex, 24 assays (Cat. No. QVT0012). It is a blue precipitating substrate with components 1 -3 used for the detection of alkaline phosphatase activity.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeSubstrate
Quantity24 Assays
Detection MethodChromogenic, Fluorescence
Unit SizeEach
Frequently asked questions (FAQs)
Does ViewRNA Tissue Assay Blue Module (Cat Nos. 19932, 19943) use the same Fast Blue reagent as used in the earlier Fast Blue Substrate products?
Yes. There was no change to the Fast Blue reagent used.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
How is ViewRNA Tissue Assay Blue Module (Cat Nos. 19932, 19943) different from the earlier Fast Blue Substrate products?
The earlier Fast Blue Substrate products (Cat Nos. QVT0506 and QVT0507) provided only the Blue Buffer and Blue Reagents 1, 2, and 3. The ViewRNA Tissue Assay Blue Module (Cat Nos. 19932, 19943) includes these same components plus a Label Probe 6-AP and a stop reagent.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.