Fast Blue Substrate (20 mL Blue Buffer + 420 uL each Blue Reagents 1-3) - FAQs
Fast Blue Substrate (20 mL Blue Buffer + 420 uL each Blue Reagents 1-3) - FAQs
View additional product information for Fast Blue Substrate (20 mL Blue Buffer + 420 uL each Blue Reagents 1-3) - FAQs (QVT0506)
2 product FAQs found
Does ViewRNA Tissue Assay Blue Module (Cat Nos. 19932, 19943) use the same Fast Blue reagent as used in the earlier Fast Blue Substrate products?
Yes. There was no change to the Fast Blue reagent used.
How is ViewRNA Tissue Assay Blue Module (Cat Nos. 19932, 19943) different from the earlier Fast Blue Substrate products?
The earlier Fast Blue Substrate products (Cat Nos. QVT0506 and QVT0507) provided only the Blue Buffer and Blue Reagents 1, 2, and 3. The ViewRNA Tissue Assay Blue Module (Cat Nos. 19932, 19943) includes these same components plus a Label Probe 6-AP and a stop reagent.