ViewRNA Tissue Assay Fluorescence Kits
Thermo Scientific™

ViewRNA Tissue Assay Fluorescence Kits

ViewRNA Tissue assays are RNA in situ hybridization (ISH) assays for the reliable detection of up to four RNA targets within tissue sections and cultured cells. Visualize any RNA in any tissue/cell with single-copy sensitivity.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeColorQuantity
QVT4646BTissue Alexa Fluor 546 ModuleOrange (TRITC)96 Slides
QVT0700Tissue Fluor. 4 Plex Assay KitGreen, Orange, Deep Red, NIR24 Slides
QVT4700Tissue Fluor. 4 Plex Assay KitGreen, Orange, Deep Red, NIR96 Slides
QVT0800Tissue Fluor. 4 Plex Assay KitGreen, Red, Deep Red, NIR24 Slides
QVT4800Tissue Fluor. 4 Plex Assay KitGreen, Red, Deep Red, NIR96 Slides
QVT0600CTissue Fluor. 1 Plex AssayDeep Red (Cy5)24 Slides
QVT4600CTissue Fluor. 1 Plex AssayDeep Red (Cy5)96 Slides
QVT0688BTissue Alexa Fluor 488 ModuleGreen (FITC)24 Slides
QVT4688BTissue Alexa Fluor 488 ModuleGreen (FITC)96 Slides
QVT0646BTissue Alexa Fluor 546 ModuleOrange (TRITC)24 Slides
QVT0694BTissue Alexa Fluor 594 ModuleRed (Texas Red)24 Slides
QVT4694BTissue Alexa Fluor 594 ModuleRed (Texas Red)96 Slides
QVT0640BTissue Alexa Fluor 750 ModuleNear Infrared (Cy7)24 Slides
QVT4640BTissue Alexa Fluor 750 ModuleNear Infrared (Cy7)96 Slides
Catalog number QVT4646B
Price (USD)
1,085.65
Online exclusive
1,292.00
Save 206.35
Each 1
Product Type:
Tissue Alexa Fluor 546 Module
Color:
Orange (TRITC)
Quantity:
96 Slides
Price (USD)
1,085.65
Online exclusive
1,292.00
Save 206.35
Each 1
ViewRNA Tissue Assay Fluorescence kits enable an in situ hybridization (ISH) method for the simultaneous detection of one to four RNA targets (mRNA, non-coding RNA, or microRNA) at single-copy sensitivity. These kits combine bright Alexa Fluor 488, Alexa Fluor 594, Alexa Fluor 546, Alexa Fluor 647, and Alexa Fluor 750 dyes that are visualized using fluorescence microscopes or high-content imagers. ViewRNA Tissue assay uses proprietary chemistry for the target-specific probe sets (RNA FISH probes) and branch DNA (bDNA) signal amplification for detection of specific signal.

With ViewRNA Tissue assays, you can achieve:

  • Robust, single RNA molecule detection in FFPE tissue sections, cryo-preserved sections, and cultured cells
  • Accuracy and specificity inherent to bDNA amplification technologies
  • Simultaneous detection of up to four RNA targets, including mRNA, noncoding RNA, and microRNA

Note: A ViewRNA probe set is required and can be purchased separately

ViewRNA Tissue Assay Fluorescence kits, when combined with a ViewRNA Tissue probe set (purchased separately), enables the detection of a single RNA target in tissues/cells using Alexa Fluor dyes. ViewRNA offers 6500 catalog probe sets. Any more probe sets can be designed at no extra cost.

To meet various experimental needs, the ViewRNA Tissue assay is provided in various kits and modules.

ViewRNA Tissue Fluorescence 4-Plex Assay Kit (green, orange, deep red, and near infrared)

With this kit four RNA targets can be detected using ViewRNA probe sets type 1, 4, 6, and 10 (ViewRNA probe sets are sold separately). This kit utilizes Alexa Fluor 546 for Type 1 probes, Alexa Fluor 488 for Type 4 probes, Alexa Fluor 647 for Type 6 probes, and Alexa Fluor 750 for Type 10 probes. RNA visualization requires FITC/GFP, TRITC/RFP, CY5/Deep Red, and Cy7/NIR microscope filter sets. An additional DAPI filter set is needed for nucleus counterstain with DAPI.

ViewRNA Tissue Fluorescence 4-Plex Assay Kit (green, red, deep red, and near infrared)

With this kit four RNA targets can be detected using ViewRNA probe sets type 1, 4, 6, and 10 (ViewRNA probe sets are sold separately). This kit utilizes Alexa Fluor 594 for Type 1 probes, Alexa Fluor 488 for Type 4 probes, Alexa Fluor 647 for Type 6 probes, and Alexa Fluor 750 for Type 10 probes. RNA visualization requires FITC/GFP, Texas Red, CY5/Deep Red, and Cy7/NIR microscope filter sets. An additional DAPI filter set is needed for nucleus counterstain with DAPI.

ViewRNA Tissue Fluorescence 1-Plex Assay Core Kit, with Alexa Fluor 647 (Type 6) label probe

With this kit, one RNA target can be detected using a Type 6 ViewRNA probe set (ViewRNA probe sets are sold separately). Up to four RNA targets can be detected when combined with ViewRNA Tissue modules (sold separately). RNA visualization requires a Cy5/Deep Red microscope filter set. An additional DAPI filter set is needed for nucleus counterstain with DAPI.

ViewRNA Tissue Alexa Fluor 488 (Type 4) Module

When combined with the ViewRNA Tissue Fluorescence 1-Plex Assay Core Kit (Cat. No. QVT0600) this module can detect an additional RNA target utilizing a Type 4 ViewRNA probe set. This module can be combined with other modules for additional RNA target detection. RNA visualization requires a FITC/GFP filter set.

ViewRNA Tissue Alexa Fluor 546 (Type 1) Module

When combined with the ViewRNA Tissue Fluorescence 1-Plex Assay Core Kit (Cat. No. QVT0600) this module can detect an additional RNA target utilizing a Type 1 ViewRNA probe set. This module can be combined with other modules for additional RNA target detection. RNA visualization requires a TRITC/RFP filter set.

Note: The ViewRNA Tissue Alexa Fluor 546 (Type 1) Module cannot be combined with the ViewRNA Tissue Alexa Fluor 594 (Type 1) Module because both use a Type 1 probe set and will spectrally overlap in most fluorescence microscopes.

ViewRNA Tissue Alexa Fluor 594 (Type 1) Module

When combined with the ViewRNA Tissue Fluorescence 1-Plex Assay Core Kit (Cat. No. QVT0600) this module can detect an additional RNA target utilizing a Type 1 ViewRNA probe set. This module can be combined with other modules for additional RNA target detection. RNA visualization requires a Texas Red filter set.

Note: The ViewRNA Tissue Alexa Fluor 594 (Type 1) Module cannot be combined with the ViewRNA Tissue Alexa Fluor 546 (Type 1) Module because both use a Type 1 probe set and will spectrally overlap in most fluorescence microscopes.

ViewRNA Tissue Alexa Fluor 750 (Type 10) Module

When combined with ViewRNA Tissue Fluorescence 1-Plex Assay Core Kit (Cat. No. QVT0600) this module can detect an additional RNA target utilizing a Type 10 ViewRNA probe set. This module can be combined with other modules for additional RNA target detection. RNA visualization requires a CY7/NIR filter set.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorOrange (TRITC)
Excitation/Emission556/573 nm
For Use With (Application)RNA-FISH, In Situ Hybridization (ISH)
Labeling MethodViewRNA Type 1
Product LineViewRNA
Product TypeTissue Alexa Fluor 546 Module
Quantity96 Slides
Detection MethodFluorescence
Final Product TypeDNA
FormatKit
Labeling TargetRNA (General)
Label or DyeAlexa Fluor 546
Sample TypeTissue (FFPE), Tissue (CRYO), Cell Culture Cells
Unit SizeEach 1

Frequently asked questions (FAQs)

How do ViewRNA assays compare to RNAScope assays?

ViewRNA and RNAScope technologies rely on the same signal amplification strategy - branched DNA amplification. Historically, both ViewRNA and RNAScope technologies originated from the same company, Panomics. The assays are expected to yield similar sensitivity and resolution, however each technology relies on its own set of proprietary reagents and probe set designs. Hence, the assays are not considered interchangeable or compatible. ViewRNA probe sets are not tested for RNAScope assaya and vice versa.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

How is the signal amplified in ViewRNA assays?

The ViewRNA technology relies on branched DNA signal amplification strategy. Target probes complementary to the target transcript sequence are further hybridized with pre-amplifier, amplifier and label probes that consist of branched DNA, and form 'tree branches' that allow numerous label probes to attach. This approach allows higher signal amplification compared to traditional ISH techniques.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Where can I find general information about ViewRNA ISH Assays?

For general information about ViewRNA ISH Assays, please go to this page (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cellular-imaging/in-situ-hybridization-ish/rna-fish/viewrna-assays.html).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

For ViewRNA assays, can I use the same set of wash solutions for all samples, including the positive and negative controls?

We do not recommend doing this. The negative control should be processed and washed separately from the rest of the samples. This is because the negative control does not contain any target probe sets and only the amplification reagents are added to it. If experimental samples are washed in the same beaker of wash solutions as the negative control, any unbound target probes that wash away can carry over to the negative control sample and cause unexpected positive signal (that will also appear to be very specific).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Is there a video on the ViewRNA tissue assay workflow that I can watch?

Yes, we do have a video that will help provide hands-on guidance on how to proceed with the protocol. Please find the link to the online protocol and video (https://www.thermofisher.com/us/en/home/references/protocols/cell-and-tissue-analysis/protocols/viewrna-tissue-fluorescence-protocol.html).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.