ATP Solution (100 mM)
Thermo Scientific™

ATP Solution (100 mM)

Thermo Scientific™ ATP (adenosine 5'-triphosphate) is an extremely stable nucleotide, and it is supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH.
Have Questions?
Catalog NumberQuantity
R04410.25 mL
Catalog number R0441
Price (KRW)
53,000
Online offer
Ends: 31-Dec-2026
58,000
Save 5,000
Each
Quantity:
0.25 mL
Request bulk or custom format
Price (KRW)
53,000
Online offer
Ends: 31-Dec-2026
58,000
Save 5,000
Each

Thermo Scientific™ ATP (adenosine 5'-triphosphate) is an extremely stable nucleotide, and it is supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH. This nucleotide is used in different molecular biology applications, and it has greater than 99% purity.

Features

  • Greater than 99% purity confirmed by HPLC
  • Functionally tested in in vitro transcription
  • Stable for two years at -20°C and after multiple freeze-thaw cycles

Applications

  • in vitro transcription
  • aRNA synthesis
  • siRNA synthesis
  • RNA amplification
  • Ligation
  • Phosphorylation

For Research Use Only. Not for use in diagnostic procedures.

Specifications
ConjugateUnconjugated
For Use With (Application)in vitro Transcription, RNA Amplification
Product TypeATP Solution
Quantity0.25 mL
Shipping ConditionDry Ice
Concentration100 mM
FormLiquid
Unit SizeEach
Contents & Storage
ATP Solution, 100 mM (0.25 mL)

Store at –20°C.