MassRuler DNA Loading Dye (6X)
Thermo Scientific™

MassRuler DNA Loading Dye (6X)

Thermo Scientific 6X MassRuler DNA Loading Dye is used to prepare DNA markers and samples for loading on agarose orRead more
Have Questions?
Catalog NumberQuantity
R06215 x 1 mL
Catalog number R0621
Price (USD)
51.65
Online exclusive
61.00
Save 9.35
Each
Quantity:
5 x 1 mL
Request bulk or custom format
Price (USD)
51.65
Online exclusive
61.00
Save 9.35
Each
Thermo Scientific 6X MassRuler DNA Loading Dye is used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It contains one dye (bromophenol blue) for visual tracking of DNA migration during electrophoresis. The presence of glycerol ensures that the DNA in the ladder and sample forms a layer at the bottom of the well. The EDTA included in the solutions binds divalent metal ions and inhibits metal-dependent nucleases.

6X MassRuler DNA Loading Dye is used for conventional DNA electrophoresis.

Highlights

• One-color tracking of DNA migration during DNA electrophoresis
• No DNA masking during gel exposure to UV light
• EDTA binds divalent metal ions and inhibits metal dependent nucleases

Applications

• Analysis of large DNA molecules
• Preparation of DNA ladders, markers, and samples for loading on agarose or polyacrylamide gels
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gel CompatibilityAcrylamide Gels, Agarose Gels
Label or DyeBromophenol Blue
Product LineMassRuler
Product TypeDNA Loading Dye
Quantity5 x 1 mL
Concentration6X
Unit SizeEach