What can I use as a substitution for the discontinued RapidRun Loading Dye?
We recommend using the Orange DNA Loading Dy (6X) (Cat. No. R0631) as a substitute for the RapidRun Loading Dye.
Search
View additional product information for Orange DNA Loading Dye (6X) - FAQs (R0631)
1 product FAQs found