ReadyLabel™ Antibody Labeling Kits
ReadyLabel™ Antibody Labeling Kits
ReadyLabel™ Antibody Labeling Kits
ReadyLabel™ Antibody Labeling Kits
ReadyLabel™ Antibody Labeling Kits
Invitrogen™

ReadyLabel™ Antibody Labeling Kits

ReadyLabel Antibody Labeling Kits enable fast, convenient labeling of off-the-shelf antibodies and cell culture supernatants without prior purification of the antibody solution.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityLabel or Dye
R107245 x 20 μgAlexa Fluor Plus 750
R107095 x 20 μg of IgGAlexa Fluor 488
R107125 x 100 μg of IgGAlexa Fluor 488
R107105 x 20 μg of IgGAlexa Fluor 647
R107135 x 100 μg of IgGAlexa Fluor 647
R107115 x 20 μg of IgGBiotin
R107145 x 100 μg of IgGBiotin
R107015 x 20 μg of IgGNone
R107025 x 100 μg of IgGNone
R107205 x 20 μgAlexa Fluor 514
R107215 x 20 μgAlexa Fluor Plus 555
R107225 x 20 μgAlexa Fluor 594
R107235 x 20 μgAlexa Fluor 700
R107255 x 20 μgAlexa Fluor 420
Catalog number R10724
Price (USD)
618.00
5 x 20 μg
Quantity:
5 x 20 μg
Label or Dye:
Alexa Fluor Plus 750
Price (USD)
618.00
5 x 20 μg
ReadyLabel Antibody Labeling Kits enable fast, convenient labeling of off-the-shelf antibodies and cell culture supernatants without prior purification of the antibody solution. Each kit contains everything needed to perform 5 labeling reactions and will produce purified antibody conjugates, without the purification step.

Each ReadyLabel Antibody Labeling Kit, 100 μg contains everything needed to perform 5 labeling reactions and will produce purified antibody conjugates, without the purification step. The kit can be used to label antibodies in solutions containing BSA or other stabilizing proteins and antibodies in a primary amine-containing buffer such as Tris or glycine, or even cell culture supernatants, without prior purification of the antibody-containing solution. A reactive dye moiety efficiently binds to primary amines of antibodies to form stable, covalently bound antibody-dye conjugates.

Alexa Fluor dyes produce antibody conjugates that are typically brighter with better photostability and lower pH sensitivity relative to traditional dyes (e.g., FITC). Unlike other dyes, the fluorescence of Alexa Fluor conjugates are insensitive to pH between 4 and 10. Alexa Fluor-labeled antibodies can be used for applications including microscopy, flow cytometry, western blotting, ELISA, and indirect FISH.

ReadyLabel Antibody Labeling Kits offer the following benefits:

  • Convenience—Labeling takes one hour, with less than 20 minutes of hands-on time
  • Versatility—Labelling can be performed on IgG antibodies, regardless of species, isotype, or storage buffer
  • Customization—ReadyLabel Flex Antibody Labeling Kits are designed for use with a wide range of amine-reactive labels including Alexa Fluor dyes
For Research Use Only.
Specifications
Detection MethodFluorescence
For Use With (Application)Antibody Production and Labeling
For Use With (Equipment)Flow Cytometer, Fluorescence Microscope
Product LineReadyLabel
Product TypeAntibody Labeling Kit
Quantity5 x 20 μg
Chemical ReactivityAmine
Labeling TargetIgG
Label or DyeAlexa Fluor Plus 750
Unit Size5 x 20 μg