Resorufin β-D-Galactopyranoside - FAQs
Resorufin β-D-Galactopyranoside - FAQs
View additional product information for Resorufin β-D-Galactopyranoside - FAQs (R1159)
1 product FAQs found
What is the excitation/emission maxima of resorufin?
The excitation/emission maxima is 563/587 nm.