Resazurin, Sodium Salt
Invitrogen™

Resazurin, Sodium Salt

Resazurin, Sodium Salt

Have Questions?
Catalog NumberQuantity
R1220410 mg
Catalog number R12204
Price (MXN)
4,196.65
Each
Quantity:
10 mg
Price (MXN)
4,196.65
Each

The non-fluorescent resazurin can be used as a fluorogenic oxidation-reduction indicator in a variety of cells, including bacteria, yeast and eukaryotes by flow cytometry, fluorescence microscopy and high-throughput screening. The red-fluorescent product, resorufin exhibits absorption/emission maxima ∼575/585 nm.

The non-fluorescent resazurin can be used as a fluorogenic oxidation-reduction indicator in a variety of cells, including bacteria, yeast and eukaryotes by flow cytometry, fluorescence microscopy and high-throughput screening. The red-fluorescent product, resorufin exhibits absorption/emission maxima ∼575/585 nm.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionResazurin, Sodium Salt
Detection MethodFluorescence
Dye TypeResazurin
Emission Wavelength585
Excitation Wavelength575 nm
FormSolid
Quantity10 mg
Shipping ConditionRoom Temperature.
ColorRed
For Use With (Application)Cell Viability
For Use With (Equipment)Microplate Reader
Product LineInvitrogen
Product TypeResazurin
Unit SizeEach
Contents & Storage
Store at room temperature and protect from light.

Frequently asked questions (FAQs)

How do alamarBlue reagent and PrestoBlue reagent differ from resazurin and C12-resazurin?

alamarBlue reagent and PrestoBlue reagent contain resazurin in a proprietary stabilizing formulation that allows for a convenient “mix, incubate, and read” protocol. PrestoBlue reagent is an improvement in the formulation of alamarBlue reagent that allows for much faster staining (typically 10 minutes vs. 1-4 hours to obtain a similar signal and sensitivity). C12-resazurin is a derivative of resazurin that has better cellular retention and thus allows for analysis on a flow cytometer and multiplexing with viability indicators and other biomarkers.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (305)

Citations & References
Abstract
Rapid screening of natural products for antimycobacterial activity by using luciferase-expressing strains of Mycobacterium bovis BCG and Mycobacterium intracellulare.
Authors:Shawar RM,Humble DJ,Van Dalfsen JM,Stover CK,Hickey MJ,Steele S,Mitscher LA,Baker W
Journal:Antimicrobial agents and chemotherapy
PubMed ID:9055994
The object of this study was to investigate the ability of a rapid luciferase assay to detect antimycobacterial activity in plant extracts. Recombinant strains of Mycobacterium bovis BCG (rBCG) and Mycobacterium intracellulare expressing firefly luciferase were used as the test organisms. Assays were conducted in a 96-well minitube format under ... More
Inhibition of growth and sensitization to cisplatin-mediated killing of ovarian cancer cells by polyphenolic chemopreventive agents.
Authors:Chan MM, Fong D, Soprano KJ, Holmes WF, Heverling H
Journal:J Cell Physiol
PubMed ID:12447990
'The polyphenolic compounds curcumin and quercetin increased sensitivity of ovarian cancer cells (CAOV3 and SKOV3) to cisplatin. The effect was obtained when the compounds were added simultaneously with cisplatin, as well as when they were added 24 h before. High serum levels of certain cytokines, for example interleukin-6 (IL-6), have ... More
Mechanism of target cell recognition by natural killer cells: characterization of a novel triggering molecule restricted to CD3- large granular lymphocytes.
Authors:Frey JL, Bino T, Kantor RR, Segal DM, Giardina SL, Roder J, Anderson S, Ortaldo JR
Journal:J Exp Med
PubMed ID:1720812
'In an attempt to identify a molecule in target recognition by CD3- large granular lymphocytes (LGL), we have generated a rabbit antiidiotypic (anti-ID) serum against a monoclonal antibody (mAb 36) that reacted with the cell membrane of K562. Flow cytometry analysis demonstrated that the anti-ID serum bound selectively to CD3- ... More
Functional in vivo MHC class II loading by endogenously synthesized glycoprotein during viral infection.
Authors:Oxenius A, Bachmann MF, Mathis D, Benoist C, Zinkernagel RM, Hengartner H
Journal:J Immunol
PubMed ID:9190921
'MHC class II presentation of antigenic peptides derived from soluble proteins is usually preceded by antigenic uptake via (nonreceptor-mediated) endocytosis by professional APCs, followed by processing in endosomal compartments. Although in vitro alternative pathways for MHC class II loading have been described for certain intracellularly synthesized proteins, the importance of ... More
Cell proliferation enhances entry of Listeria monocytogenes into intestinal epithelial cells by two proliferation-dependent entry pathways.
Authors:Velge P, Bottreau E, Van-Langendonck N, Kaeffer B
Journal:J Med Microbiol
PubMed ID:9511817
'Bacterial entry into intestinal host cells is the result of a fairly sophisticated manipulation of host cell machinery by the pathogens. To study further the potential cell target of Listeria spp., the in-vitro entry of L. monocytogenes strains into intestinal cells was examined in relation to the metabolism, proliferation and ... More