Product Image
Thermo Scientific™

Maxima First Strand cDNA Synthesis Kit Reaction Mix (5X)

Thermo Scientific Maxima 5X Reaction Mix is a component of the Maxima First Strand cDNA Synthesis Kits for RT-qPCR (K1671/K1672/K1641/K1642)Read more
Have Questions?
Catalog NumberQuantity
R13620.8 mL
Catalog number R1362
Price (USD)
365.00
Each
Quantity:
0.8 mL
Price (USD)
365.00
Each

Thermo Scientific Maxima 5X Reaction Mix is a component of the Maxima First Strand cDNA Synthesis Kits for RT-qPCR (K1671/K1672/K1641/K1642) made available here for separate purchase. The 5X Reaction Mix is comprised of reaction buffer, dNTPs, and oligo (dT)18 and random hexamer primers.

Oligo (dT)18 is single-stranded deoxythymine sequence used for priming reactions catalysed by reverse transcriptase.

Related products
Maxima First Strand cDNA Synthesis Kit for RT-qPCR, with dsDNase
Maxima First Strand cDNA Synthesis Kit for RT-qPCR

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeFirst-Strand cDNA
For Use WithMaxima First Strand cDNA Synthesis Kit for RT-qPCR
FormatKit
No. of Reactions200 Reactions
Quantity0.8 mL
Reaction FormatPremixed Components
Reagent TypeReverse Transcription
Shipping ConditionDry Ice
Starting MaterialRNA
Concentration5X
Unit SizeEach
Contents & Storage

• 5X Reaction Mix with oligo(dT)18 and random hexamer primers

Store at –20°C.