Yellow Sample Buffer (40X) - FAQs
Yellow Sample Buffer (40X) - FAQs
View additional product information for Yellow Sample Buffer (40X) - FAQs (R1381)
1 product FAQs found
Does the yellow dye from the Yellow Sample Buffer provided with Luminaris Color and DyNAmo Flash qPCR Master Mixes influence qPCR performance?
The addition of yellow dye from the Yellow Sample Buffer to qPCR reaction has no influence on the efficiency, specificity, sensitivity or Cq values. Using the Yellow Sample Buffer is optional, but the dye helps to see in which wells the sample has been already added.