Remel™ Haemophilus Identification Test Kit
Thermo Scientific™
Remel™ Haemophilus Identification Test Kit
Use for determination of growth requirements and porphyrin production by Haemophilus spp.
Have Questions?
Catalog NumberQuantity
R2112525 Tests/Kit
Catalog number R21125
Price (USD)
-
Quantity:
25 Tests/Kit
Thermo Scientific™ Remel Haemophilus Identification Test Kit is used for determination of growth requirements and porphyrin production by Haemophilus spp.
Includes:
Hemin (X factor) discs (H); and V Factor with ALA discs (VP)
Specifications
DescriptionHaemophilus Identification Test Kit
Packaging Type1 Kit
Quantity25 Tests/Kit
TypeHaemophilis Testing Kit
Detectable AnalytesHaemophilus species
Unit SizeEach