NucRed™ Live 647 ReadyProbes™ Reagent - FAQs
NucRed™ Live 647 ReadyProbes™ Reagent - FAQs
View additional product information for NucRed™ Live 647 ReadyProbes™ Reagent - FAQs (R37106)
2 product FAQs found
How do I get rid of precipitates in NucRed Live 647 ReadyProbes Reagent (Cat. No. R37106)?
Briefly warm NucRed Live 647 ReadyProbes Reagent to 37 degrees C and vortex.
Can I use the ReadyProbes reagents for flow cytometry?
This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.