Propidium Iodide ReadyProbes™ Reagent - FAQs
Propidium Iodide ReadyProbes™ Reagent - FAQs
View additional product information for Propidium Iodide ReadyProbes™ Reagent - FAQs (R37108)
2 product FAQs found
What is the volume for one drop from the ReadyProbes dropper bottles?
The volume for one drop from the ReadyProbes dropper bottles is approximately 41 µL.
Can I use the ReadyProbes reagents for flow cytometry?
This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.